Skip to Content
MilliporeSigma

Skip To

MR-024

EmbryoMax® FHM Mouse Embryo Media

(1X), liquid, with Phenol Red

Synonym(s):

EmbryoMax Media, FHM Mouse Embryo Media, Mouse Embryo Media

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
50 mL
Check Cart for Availability
$42.90

About This Item

UNSPSC Code:
12352207
NACRES:
NA.71
eCl@ss:
32160801

$42.90


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

liquid

manufacturer/tradename

Specialty Media, EmbryoMax®

technique(s)

cell culture | embryo: suitable, cell culture | stem cell: suitable

input

sample type: mouse embryo(s)

General description

EmbryoMax® FHM Mouse Embryo Media is used to enable embryo collection, manipulation, and transfer techniques. offers a range of mouse embryo media and reagents for your research needs.

Application

EmbryoMax® FHM Mouse Embryo Media has been used to recover mouse fertilized eggs or embryos from the oviduct.[1][2][3]

Preparation Note

Can be stored at -20°C up to the expiration date on the bottle. Once thawed it should be used within 2 weeks.

Legal Information

EmbryoMax is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MR-025MR-056MR-070
form

liquid

form

liquid

form

liquid

form

liquid

technique(s)

cell culture | embryo: suitable, cell culture | stem cell: suitable

technique(s)

cell culture | embryo: suitable, cell culture | stem cell: suitable

technique(s)

cell culture | stem cell: suitable

technique(s)

cell culture | embryo: suitable, cell culture | stem cell: suitable

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300

manufacturer/tradename

Specialty Media, EmbryoMax®

manufacturer/tradename

Specialty Media, EmbryoMax®

manufacturer/tradename

Specialty Media, EmbryoMax®

manufacturer/tradename

Specialty Media, EmbryoMax®

input

sample type: mouse embryo(s)

input

sample type: mouse embryo(s)

input

sample type: mouse embryo(s)

input

sample type: mouse embryo(s)


Still not finding the right product?

Explore all of our products under EmbryoMax® FHM Mouse Embryo Media


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters