Skip to Content

System maintenance is scheduled for 11 Sep, 08:00 PM - 12 Sep, 00:00 AM CST [12 Sep 03:00 AM to 07:00 AM CET] and will affect some web and offline transactions, including quotes, price checks, and order status inquiries. We apologize for any inconvenience.

MilliporeSigma

Skip To

MR-015

M2 medium

EmbryoMax M2 Medium (1X), Liquid, with Phenol Red

Synonym(s):

M2 Embryo Media

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352207
NACRES:
NA.71
eCl@ss:
32160801
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

liquid

manufacturer/tradename

Specialty Media, EmbryoMax®

technique(s)

cell culture | embryo: suitable, cell culture | stem cell: suitable

input

sample type: mouse embryo(s)

Application

EmbryoMax M2 Medium (1X), Liquid, with Phenol Red

Preparation Note

Can be stored at -20°C up to the expiration date on the bottle. Once thawed it should be used within 2 weeks.

Frozen media can be placed in a refrigerator (2°C to 8°C) to thaw overnight. The thawed media should be swirled gently to mix the media, and warmed to 37°C prior to use. Altematively, place frozen medium into a 37°C water bath until thawed completely. It is best to gently swirl the medium during thawing to accelerate the process, and mix the medium. Do not allow the thawed medium to remain in the 37°C water bath for extended periods after it has thawed completely, and warmed to 37°C.

Once thawed the medium has a shelf life of 2 weeks, and must be stored at 2°C to 8°C between uses. Do not re-freeze the medium. Medium stored beyond this 2 week period may not perform optimally.

Note 1:


The "Last Thaw Date" on the label indicates the last day in the shelf life of the frozen media. Media stored frozen beyond this date may not perform optimally. Media are best used prior to the "Last Thaw Date" however, once the "Last Thaw Date" has been reached, the media should be thawed, stored at 2°C to 8°C, and used within 2 weeks.

Note 2:


Although the pH of these media are correct when thawed, the pH of the sodium bicarbonate buffered formulations will rise as the media are exposed to air. This rise in pH will increase with the frequency of opening the bottle, and is particularly rapid when drops of medium placed on a dish for embryo culture are exposed to air. Thus, it is recommended that the drops of medium (placed under light mineral oil) are pre-equilibrated 6 hours to overnight in a humidified incubator at 37°C, in a 5% C02 + 95% Air atmosphere, prior to introducing embryos into the medium.

Legal Information

EmbryoMax is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Explore all of our products under M2 medium


Storage Class

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–4 of 4 Questions  
  1. What is the full product formulation for the MR-015 formulation of M2 medium?

    1 answer
    1. The fomulation of this medium is as follows:
      Component mM
      NaCl 94.62
      KCl 4.78
      KH2PO4 1.19
      MgSO4-7H2O 1.19
      CaCl-2H2O 1.71
      NaHCO3 4.15
      HEPES 20.85
      D-Glucose 5.56
      Sodium Lactate 23.28
      Sodium Pyruvate 0.33
      Bovine Serum Albumin 4.0 g/L
      Penicillin G Sodium 100 U/mL
      Streptomycin 0.05 g/L
      Phenol Red 0.01 g/L

      Helpful?

  2. Does the MR-015 formulation of M2 medium contain BSA and what concentration?

    1 answer
    1. Yes, this product contains Bovine Serum Albumin at a concentration of 4.0 g/L, equivalent to 0.4% w/v.

      Helpful?

  3. Does the MR-015 formulation of M2 medium contain Bovine Serum Albumin, if so what tissue the concentration?

    1 answer
    1. Yes, this product contains Bovine Serum Albumin at a concentration of 4.0 g/L, equivalent to 0.4% w/v.

      Helpful?

  4. Does the MR-015 formulation of M2 medium contain antibiotics?

    1 answer
    1. The MR-015 formulation of M2 medium does contain antibiotics.

      Helpful?

Reviews

No rating value

Active Filters