Skip to Content
MilliporeSigma

MPBD

mPAGE®Buffer Dam

for mPAGE® Mini Gel Tank

Synonym(s):

Buffer adapter

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.56
UNSPSC Code:
41105300
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

electrophoresis grade

description

For use in mini gel electrophoresis tanks when running an odd number of mPAGE® Precast Gels

packaging

pack of 1 packs

manufacturer/tradename

Millipore

technique(s)

SDS-PAGE: suitable, electrophoresis: suitable, western blot: suitable

compatibility

for use with mPAGE® Mini Gel Tank (smooth side of the gasket visible)

General description

mPAGE® Buffer Dam for use mini gel tanks when running an odd number of mPAGE® Precast Gels.

Packaging

1 each

Legal Information

MPAGE is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under mPAGE®Buffer Dam




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
MPBD04054839761683