Skip to Content
MilliporeSigma

Skip To

MP12W12

mPAGE® 12% Bis-Tris Precast Gels

12% Bis-Tris, 12 well, 1mm

Synonym(s):

Bis-Tris precast gels, precast gels for SDS-PAGE, precast polyacrylamide gels

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41105320
NACRES:
NA.56
Cassette w × l:
10 cm × 8 cm
Gel thickness:
1 mm
Well maximum volume:
60 μL
Compatibility:
mPAGE® MES or MOPS Running Buffer; mPAGE® Mini Gel System
Packaging:
pkg of 10 gels
Shelf life:
18 mo.
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

mPAGE® 12% Bis-Tris Precast Gel, L × W × wells 10 cm × 8 cm × 12 well

shelf life

18 mo.

packaging

pkg of 10 gels

L × W × wells

10 cm × 8 cm × 12 well

cassette W × L

10 cm × 8 cm

gel thickness

1 mm

well maximum volume

60 μL

well wells

12 wells

compatibility

mPAGE® MES or MOPS Running Buffer; mPAGE® Mini Gel System

storage temp.

2-8°C

General description

mPAGE 12% Bis-Tris Precast Gel, 10x8 cm
  • Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Application

mPAGE 12% bis-tris gels for high resolution protein separation

  • Load up to 60 μl sample / well
  • Typical run time 30 min (MES, 180V) or 34 min (MOPS, 200V)
  • Reverse gasket when using with Bio-Rad Mini-PROTEAN systems
  • Use adapter (MPTA) with XCell SureLock and other compatible systems

Legal Information

MPAGE is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MP12W10MP10W12MP12W15
description

L × W × wells 10 cm × 8 cm × 12 well

description

L × W × wells 10 cm × 8 cm × 10 well

description

L × W × wells 10 cm × 8 cm × 12 well

description

L × W × wells 10 cm × 8 cm × 15 well

compatibility

mPAGE® MES or MOPS Running Buffer; mPAGE® Mini Gel System

compatibility

mPAGE® MES or MOPS Running Buffer; mPAGE® Mini Gel System

compatibility

mPAGE® MES or MOPS Running Buffer; mPAGE® Mini Gel System

compatibility

mPAGE® MES or MOPS Running Buffer; mPAGE® Mini Gel System

cassette W × L

10 cm × 8 cm

cassette W × L

10 μL × 8 μL

cassette W × L

10 cm × 8 cm

cassette W × L

10 cm × 8 cm

well maximum volume

60 μL

well maximum volume

80 μL

well maximum volume

60 μL

well maximum volume

40 μL

gel thickness

1 mm

gel thickness

1 mm

gel thickness

1 mm

gel thickness

1 mm

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

shelf life

18 mo.

shelf life

18 mo.

shelf life

18 mo.

shelf life

18 mo.


Still not finding the right product?

Explore all of our products under mPAGE® 12% Bis-Tris Precast Gel


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. Dear all, I would like to ask for the expiry date of this product (MP12W12 Millipore mPAGE® 12% Bis-Tris vorgegossenes Gel). Thanks, best regards, Martin

    1 answer
    1. The expiration date will be listed on the lot specific Certificate of Analysis. Please navigate to the link below to view the current documentation available:

      https://www.emdmillipore.com/product/mPAGE-120-0-Bis-Tris-Precast-Gel-10x8-cm-12-well,MM_NF-MP12W12#documentation

      Helpful?

Reviews

No rating value

Active Filters