Skip to Content
MilliporeSigma

Skip To

MP0DSYS3A

Process Scale Holders

for use with Pod Filters, 316L stainless steel

Synonym(s):

Process Scale Holders 3 Racks

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

eCl@ss:
32031690
UNSPSC Code:
23151806
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

material

316L stainless steel , silicone seal

description

3-rack holder; for Pod configurations of 5.5 m2 to 33 m2; Gemü® valves

fitting

1.5 in. gauge port sanitary flange, 1.5 in. inlet/outlet sanitary flange

compatibility

for use with Pod Filters

Application

3-rack holder; for Pod configurations of 5.5 m2 to 33 m2; Gem® valves

Legal Information

GEM is a registered trademark of Welch Vacuum Technology

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MP0DSYS2AMP0DSYS1BMP0DSYS1N
fitting

1.5 in. gauge port sanitary flange, 1.5 in. inlet/outlet sanitary flange

fitting

1.5 in. gauge port sanitary flange, 1.5 in. inlet/outlet sanitary flange

fitting

1.5 in. gauge port sanitary flange, 1.5 in. inlet/outlet sanitary flange

fitting

1.5 in. gauge port sanitary flange, 1.5 in. inlet/outlet sanitary flange

material

316L stainless steel , silicone seal

material

316L stainless steel , silicone seal

material

316L stainless steel , silicone seal

material

316L stainless steel , silicone seal

description

3-rack holder; for Pod configurations of 5.5 m2 to 33 m2; Gemü® valves

description

2-rack holder; for Pod configurations of 5.5 m2 to 22 m2; Gemü® valves

description

1-rack holder; for Pod configurations of 5.5 m2 to 11 m2; ITT valves

description

1-rack holder; for Pod configurations of 5.5 m2 to 11 m2; ITT valves

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300

compatibility

for use with Pod Filters

compatibility

for use with Pod Filters

compatibility

for use with Pod Filters

compatibility

for use with Pod Filters


Still not finding the right product?

Explore all of our products under Process Scale Holders




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters