Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MIACSUP01

Millipore

Microfil 1-post Base w/tubing no Tulip

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

manufacturer/tradename

Microfil® with S-Pak®

Quality Level

input

sample type beverage(s)
sample type food(s)

application(s)

bioburden testing
clinical
cosmetics
environmental
food and beverages
industrial qc
water monitoring

shipped in

ambient

General description

Microfil Spare Part

Legal Information

MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany
S-PAK is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service