Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MHA00P2SM

Millipore

Yeast and Mold Selective Broth

capacity 2 mL, Ampoule for testing soft drinks and other beverages for growth of yeast and mold. Antibiotics in the medium prevent bacterial overgrowth.

Synonym(s):

YM broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106226
eCl@ss:
32160801
NACRES:
NA.74

Quality Level

sterility

sterile

form

liquid

shelf life

12 mo.

feature

ready-to-use

packaging

pack of 50 tests

capacity

2 mL

filling volume

2 mL

color

green

input

sample type beverage(s)
sample type cider
sample type food(s)
sample type beverage(s)
sample type wine
sample type soft drink
sample type beer
sample type cosmetics
sample type bottled water
sample type sport drink

pH

4.4±0.2 (25.0 °C)

application(s)

clinical
cosmetics
environmental
food and beverages
industrial qc (microbial monitoring)
microbiology

compatibility

for use with MF-method
for use with Microfil®
for use with MicropreSure®
for use with Milliflex®

shipped in

wet ice

storage temp.

2-8°C

suitability

molds
yeasts

General description

The microbiology of beverages will vary greatly depending upon the method of processing and the means of preservation. High microbial populations often indicate poor quality in raw material, unsanitary equipments or opportunity for growth in the food at some stage in the process. Yeast and Mould Selective Broth is used for the detection of fungi in routine analysis of beverages using membrane filter technique.(1) This medium is used for enrichment of yeasts and moulds from populations containing bacteria. The medium is highly nutritious for the growth of yeasts and moulds. Biopeptone and yeast extract provide nitrogenous compounds and vitamin B complex. Thiamine is also a B vitamin in the medium. Dextrose acts as the energy source. Diastase is a mixture of amylolytic enzymes. Bromocresol green is the pH indicator, which is green at acidic pH (pH 4.0) while blue at pH 5.6. Potassium phosphate helps in maintaining buffering action in the medium. The low pH inhibits bacterial growth. yeast produces green, opaque large colonies. Mold colonies are green and filamentous

Application

Yeast and Mould Selective Broth is used for testing soft drinks and other beverages for growth of yeast and mold. Antibiotics in the medium prevent bacterial overgrowth in samples such as Beer, Bottled Water, Cider, Soft Drink, Sport Drink and Wine. The media finds its application in Clinical, Cosmetics, Environmental Analysis, Food and Beverage and Industrial QC.

Packaging

2 mL plastic ampoules

Specifications

Incubation Temperature: 28–32 °C
Incubation Time: 48–72 hrs

Culture Medium

Yeast and Mold Selective

Analysis Note

QC organisms
S. cerevisiae (ATCC 7754)

A.brasilliensisformerly A. niger (ATCC 16404 / WDCM 00053)
Target Microorganisms: Yeast and mold (selection by antibiotics)
Yeast produce green, opaque large colonies. Mold colonies are green and filamentous

Legal Information

MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany
MICROPRESURE is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service