Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAC143

Sigma-Aldrich

Anti-Strep-Tag Antibody, clone C23.21

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

C23.21, monoclonal

mol wt

observed mol wt ~17 kDa

purified by

using protein G

species reactivity

all

packaging

antibody small pack of 100 μL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

Unknown

UniProt accession no.

shipped in

2-8°C

target post-translational modification

unmodified

General description

Strep-Tag is a short amino acid-peptide sequence that exhibits specific binding to streptavidin. It is shown to reversibly occupy the same pocket where biotin is normally complexed. Hence, it is highly desirable for applications involving where efficient purification of fusion proteins. Use of this tag allows for easier detection of fusion proteins in various immunochemical assays. Strep-Tag is shown to be resistant to the action of proteases and can also be used in the presence of mild detergents. Clone C23.21 recognizes fusion proteins with Strep-Tag peptide sequence WSHPQFEK with picomolar affinity. It recognizes both the single and the double Strep-Tag sequence.

Specificity

Clone C23.21 is a mouse monoclonal antibody that detects Strep-Tagged proteins. It binds to WSHPQFEK sequence with picomolar affinity.

Immunogen

Recombinant ectodomain of glycoprotein E2 of GB virus (GBV-B), containing a double Strep-Tag (two Strep-Tags separated by a linker peptide)

Application

Quality Control Testing

Evaluated by Western Blotting with Strep-Tagged VHH 9.

Western Blotting Analysis (WB): A 1:500 dilution of this antibody detected Strep-Tagged VHH9.

Tested Applications

Surface plasmon resonance: A representative lot detected Strep-Tag in Surface plasmon resonance application. (Meola, A., et al. (2015). J Virol. 89(4):2170-81).

Western Blotting Analysis: A representative lot detected Strep-Tag in human brain extract and GFAP in Western Blotting applications. (Li, T., et al. (2017). Immunol Lett. 188:89-95).

Immunohistochemistry Applications: A representative lot detected Strep-Tag in mouse brain tissue sections in Immunohistochemistry applications (Li, T., et al. (2017). Immunol Lett. 188:89-95).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Strep-Tag, clone C23.21, Cat. No. MAC143, is a mouse monoclonal antibody that detects Strep-Tagged proteins and is tested for use in Immunohistochemistry, Surface plasmon resonance, and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service