Skip to Content
MilliporeSigma

MAC140

Anti-Inntag 10 (IT10) Antibody, clone IT10

clone IT10-1, from mouse

Synonym(s):

Allergen Amb t V, Allergen Ra5G, Amb t 5

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
IT10-1, monoclonal
Application:
ICC, WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

IT10-1, monoclonal

species reactivity

Ambrosia trifida

species reactivity (predicted by homology)

all

packaging

antibody small pack of 25 μg

technique(s)

immunocytochemistry: suitable, western blot: suitable

isotype

IgG1κ

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

General description

Pollen allergen Amb t 5 (UniProt: P10414; also known as Allergen Amb t V, Allergen Ra5G, Amb t 5) binds to IgE and causes allergic reaction in human. Inntag IT10 is a small protein domain of Ambrosia trifida (Giant ragweed) that displays strong structural determinants and does not generate solubility issues in vivo and in vitro. Also, it does not compromise cell fitness when overexpressed, and has no detectable functional and localization effects when fused to a variety of target proteins. Structure of the Inntag IT10 is based on a packed beta-sheet conformation.
~33.7 kDa observed; 8.06 kDa calculated. 33.7 kD observed corresponds to 29.4 kDa GFP plus 4.3 kDa IT10. Uncharacterized bands may be observed in some lysate(s).

Immunogen

His-tagged recombinant peptide corresponding to 40 amino acids from Ambrosia trifida.

Application

Anti-Inntag 10 (IT10), clone IT10 is a mouse monoclonal antibody that detects the Innocuous Inntag IT10 based on pollen allergen Abt t5 sequence and is tested for use in Immunocytochemistry and Western Blotting.
Research Category
Secondary & Control Antibodies
Western Blotting Analysis: A representative lot detected Inntag 10 (IT10) in Western Blotting applications (Georgieva, M.V., et. al. (2015). Nat Methods. 12(10):955-8).

Immunocytochemistry Analysis: A representative lot detected Inntag 10 (IT10) in Immunocytochemistry applications (Georgieva, M.V., et. al. (2015). Nat Methods. 12(10):955-8).

Biochem/physiol Actions

Clone IT10-1 detects pollen allergen Abt t 5 in HEK293 cells transfected with GFP-IT10.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in PBS without azide.

Preparation Note

Stable for 1 year at -20°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in transfected HEK293/GFP-IT10 cell lysates.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Inntag 10 (IT10) in 10 µg of transfected HEK293/GFP-IT10 cell lysates.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library