Skip to Content
MilliporeSigma
All Photos(1)

Documents

MABX1933

Sigma-Aldrich

Anti-HSA (10mg) KC

from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

monoclonal

species reactivity

human

technique(s)

ELISA: suitable
immunodiffusion: suitable

isotype

IgG1

General description

Human Serum Albumin (HSA) antibody. HSA is the most abundant protein found in blood accounting for roughly 50% of human serum proteins. HSA mainly works as a carrier for various nutrients, metabolites and xenobiotics. It interacts with plasma zinc and regulates uptake of zinc in cells. HSA plays a crucial role in drug pharmacokinetics. It also controls oncotic pressure and volume of the blood. Mutations in the gene are linked with congenital analbuminemia and familial dysalbuminemic hyperthyroxinemia

Specificity

Specific for human serum albumin (HSA). Negative for bovine albumin, mouse albumin

Immunogen

corresponding to Human Albumin.

Application

Detect Albumin using this mouse monoclonal Anti-HSA (10mg) KC, Cat. No. MABX1933-10KC, validated for use in ELISA and Lateral Flow Assay.
Research Category
Apoptosis & Cancer
Suggested antibody pairs MABX1932-1KC and MABX1933-1KC

Quality

Tested

Physical form

Format: Purified
Protein A affinity chromatography
Purified mouse monoclonal in buffer containing 50 mM NaCl, 10 mM PBS, pH 8.0, 0.1 % sodium azide (NaN₃).

Storage and Stability

2-8°C. For long-term storage, freeze at -20°C.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service