Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABT94

Sigma-Aldrich

Anti-Myomesin-2 Antibody, clone 3B9.3

clone 3B9.3, from mouse

Synonym(s):

Myomesin-2, 165 kDa connectin-associated protein, 165 kDa titin-associated protein, M-protein, Myomesin family member 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

3B9.3, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

western blot: suitable

isotype

IgG2aλ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... MYOM2(9172)

General description

Myomesin-2 (UniProt: P54296; also known as 165 kDa connectin-associated protein, 165 kDa titin-associated protein, M-protein, Myomesin family member 2) is encoded by the MYOM2 gene (Gene ID: 9172) in human. Myomesin-2 is a major component of the vertebrate myofibrillar M band and is mainly expressed in fast fibers. It binds myosin, titin, and light meromyosin in a dose-dependent manner. Its primary function is to upkeep M-band filament lattice. It has a unique N-terminal domain followed by 12 repeat domains with strong homology to either fibronectin type III or immunoglobulin C2 domains. Myomesin-2 is shown to interact with dysferlin, a protein that aids in repairing the sarcolemma when it is damaged or torn due to muscle strain and assists in myofibril stability.

Specificity

Clone 3B9.3 detects human muscle Myomesin-2. It targets an epitope with in the N-terminal half.

Immunogen

GST-tagged recombinant fragment corresponding to 154 amino acids from the N-terminal region of human Myomesin-2.

Application

Anti-Myomesin-2, clone 3B9.3, Cat. No. MABT94, is a mouse monoclonal antibody that detects human Myomesin-2 and has been tested for use in Western Blotting.
Research Category
Cell Structure

Quality

Evaluated by Western Blotting in human skeletal muscle tissue lysate.

Western Blotting Analysis: 0.05 µg/mL of this antibody detected Myomesin-2 in human skeletal muscle tissue lysate.

Target description

~165 kDa observed; 164.90 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service