Skip to Content
MilliporeSigma

MABT366

Anti-Claudin-1/CLDN1 Antibody, clone 7A5

clone 7A5, from mouse

Synonym(s):

Claudin-1, Senescence-associated epithelial membrane protein

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
7A5, monoclonal
Application:
FACS, ICC, WB
Citations:
1
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7A5, monoclonal

species reactivity

human

should not react with

mouse

technique(s)

flow cytometry: suitable, immunocytochemistry: suitable, western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

General description

Claudin-1 (UniProt O95832; also known as Senescence-associated epithelial membrane protein) is encoded by the CLDN1 (also known as CLD1, ILVASC, NISCH, SEMP1) gene (ORF UNQ481/PRO944; Gene ID 9076) in human. Tight junctions (TJs) are highly specialized membrane barriers that block the free passage of small molecules, microorganisms, and cells across the paracellular space, as well as provide a physical barrier between the apical and basolateral membrane domains of a polarized epithelial cell. Claudins are TJ proteins whose main function is to regulate the passage of ions across the paracellular space. There exist at least 27 human claudins with differential epithelial expression patterns, allowing tissue-specific paracellular transport of ions. Claudin-1 is expressed on apical and basolateral surfaces of hepatocytes in normal liver tissue. Claudin-1 is a known host factor for hepatitis C virus (HCV) entry via interaction with CD81 and cell-to-cell HCV transmission. Claudin-1 is a four-transmembrane (a.a. 8-28, 82-102, 116-136, 164-184) protein, having both its N- and C-terminal ends exposed cytoplasmically (a.a. 1-7 & 185-211).
~20 kDa observed. 22.74 kDa calculated.

Immunogen

Epitope: Second extracellular loop.
Recombinant human Claudin-1/CLDN1.

Application

Flow Cytometry Analysis: 5.0 µg/mL from a representative lot immunostained HT1080 cells expressing exogenously transfected human claudin-1/CLDN1 and Huh-7.5.1 human hepatoma cells, but not mock-transfected HT1080 cells or non-claudin-1-expressing S7-d6 cells (Courtesy of Professor Masuo Kondoh, PhD, Osaka University, Japan).
Western Blotting Analysis: 0.5 µg/mL from a representative lot detected exogenously expressed claudin-1 in CLDN1-transfected, but not mock-transfected, HT1080 cells (Courtesy of Professor Masuo Kondoh, PhD, Osaka University, Japan).
Immunocytochemistry Analysis: A representative lot immunostained the surface of Huh-7.5.1 human hepatoma cells, but not the non-claudin-1-/CLDN1-expressing S7-A cells (Fukasawa, M., et al. (2015). J. Virol. 89(9):4866-4879).
Flow Cytometry Analysis: A representative lot specifically immunostained HT1080 cells expressing exogenously transfected human claudin-1 (CLDN1), but not HT1080 cells expressing human claudin-2, -4, -5, -6, -7, or -9, nor L cells expressing mouse claudin-1 (Fukasawa, M., et al. (2015). J. Virol. 89(9):4866-4879).
Flow Cytometry Analysis: A representative lot immunostained HEK293T transfectants expressing FLAG-tagged human claudin-1/CLDN1 and human-mouse claudin-1 chimeras with the second human extracellular loop (EL2), but not chimeras with the mouse EL2. M152L, but not V155I, mutation in human EL2 abolished the immunoreactivity (Fukasawa, M., et al. (2015). J. Virol. 89(9):4866-4879).
Western Blotting Analysis: A representative lot detected FLAG-tagged human claudin-1/CLDN1 and human-mouse claudin-1 chimeras with the second human extracellular loop (EL2), but not chimeras with the mouse EL2. M152L, but not V155I, mutation in the second human extracellular loop abolished the immunoreactivity (Fukasawa, M., et al. (2015). J. Virol. 89(9):4866-4879).
ELISA Analysis: A representative lot detected claudin-1/CLDN1 immunoreactivity in 3.7% formaldehyde-fixed Huh-7.5.1 human hepatoma cells by "cell ELISA" (Fukasawa, M., et al. (2015). J. Virol. 89(9):4866-4879).
Neutralization Analysis: A representative lot inhibited HCV infection of cultured Huh-7.5.1 human hepatoma cells in vitro and of human liver-chimeric mice in vivo (Fukasawa, M., et al. (2015). J. Virol. 89(9):4866-4879).
Research Category
Cell Structure
Research Sub Category
Infectious Diseases - Viral
This Anti-Claudin-1 Antibody, clone 7A5 is validated for use in Immunocytochemistry, Western Blotting, Flow Cytometry for the detection of CLDN1.

Biochem/physiol Actions

Clone 7A5 specifically recognized human claudin-1/CLDN1 by targeting an epitope in the second extracellular loop and prevented claudin-1-dependent hepatitis C virus (HCV) entry into host cells. Clone 7A5 does not react with human claudin-2, -4, -5, -6, -7, or -9, nor mouse claudin-1 (Fukasawa, M., et al. (2015). J. Virol. 89(9):4866-4879).

Physical form

Format: Purified
Protein G purified.
Purified mouse monoclonal IgG1κ antibody in PBS without preservatives.

Preparation Note

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Evaluated by Immunocytochemistry in HepG2 cells.

Immunocytochemistry Analysis: 10 µg/mL of this antibody detected Claudin-1/CLDN1 in HepG2 cells.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library