Skip to Content
MilliporeSigma

MABT166

Anti-Tom20/Tomm20 Antibody, clone 2F8.1

clone 2F8.1, from mouse

Synonym(s):

Mitochondrial import receptor subunit TOM20 homolog, Mitochondrial 20 kDa outer membrane protein, Outer mitochondrial membrane receptor Tom20, TOM20, TOMM20

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
2F8.1, monoclonal
Application:
ICC, WB
Citations:
23
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2F8.1, monoclonal

species reactivity

human, mouse

technique(s)

immunocytochemistry: suitable, western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TOMM20(9804)

General description

Mitochondrial import receptor subunit TOM20 homolog (TOM20 or TOMM20), also known as mitochondrial 20 kDa outer membrane protein or outer mitochondrial membrane receptor Tom20, is a central component of the pre-protein translocase complex of the outer mitochondrial membrane (TOM complex) which is responsible for the recognition and translocation of cytosolically synthesized mitochondrial pre-proteins. The TOM complex consists of at least 7 different proteins (TOMM5, TOMM6, TOMM7, TOMM20, TOMM22, TOMM40 and TOMM70). TOM20 interacts with TOM22 and facilitates the movement of pre-proteins into the TOM40 translocation pore. Also interacts with APEX1.
~ 16 kDa observed

Immunogen

GST-tagged recombinant protein corresponding to human Tom20/Tomm20.

Application

Anti-Tom20/Tomm20, clone 2F8.1 detects levels of Tom20/Tomm20 proteins & has been published & validated for use in WB & ICC.
Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected Tom20/Tomm20 in A431, NIH/3T3, and HeLa cells.
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate
Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 0.2 µg/mL of this antibody detected Tom20/Tomm20 in 200 µg of HeLa cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library