Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABT1318

Sigma-Aldrich

Anti-BCL-6 Antibody, clone GI191E/A8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

antibody form

purified antibody

clone

GI191E/A8
monoclonal

purified by

using protein G

species reactivity

mouse, human, rat, human, mouse, mouse, rat

concentration

(Please refer to lot specific datasheet.)

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

UniProt accession no.

General description

pCDNA3‐BCL6 plasmid vector (full length sequence of human BCL-6).

Specificity

Clone GI191E/A8 specifically detects BCL-6 in Human, Mouse, and Rat tissues.

Application

Enzyme Immunoassay (ELISA) Analysis: A representative lot detected BCL-6 in ELISA applications (Garcia, J.F., et. al. (2006). J Histochem. Cytyochem. 54(1):31-8).

Western Blotting Analysis: A representative lot detected BCL-6 in Western Blotting applications (Garcia, J.F., et. al. (2006). J Histochem. Cytyochem. 54(1):31-8).

Immunohistochemistry (Paraffin) Analysis: A representative lot detected BCL-6 in Immunohistochemistry applications (Garcia, J.F., et. al. (2006). J Histochem. Cytyochem. 54(1):31-8; Nan-Cha, S.H., et. al. (2009). Mod Pathol. 22(8):1006-15).

Quality

Evaluated by Immunohistochemistry (Paraffin) in human tonsil tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:2,000 dilution of this antibody detected BCL-6 in human tonsil tissue sections.

Physical properties

78.85 kDa calculated.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Mouse, Human, Rat.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service