Skip to Content
MilliporeSigma

MABS65

Anti-Insulin Receptor Antibody, beta subunit, clone CT-3

clone CT-3, from mouse

Synonym(s):

CD220 antigen, insulin receptor

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
CT-3, monoclonal
Application:
IHC, WB
Citations:
7
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

CT-3, monoclonal

species reactivity

mouse, rat (demonstrated by independent lab), human

technique(s)

immunohistochemistry: suitable, western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... INSR(3643)
mouse ... Insr(16337)
rat ... Insr(24954)

General description

95 kDa
The insulin receptor is a tyrosine kinase receptor that when bound to insulin, initiates multiple signal transduction pathways, including JNK, PI 3-kinase, Akt and PKC. Pharmacological intervention of these Insulin R-dependent pathways is of great interest for the treatment of insulin resistance, obesity and diabetes. The Insulin Receptor (IR) is synthesized as a single polypeptide, which is subsequently cleaved to generate an extracellular α-chain and a transmembrane and intracellular β-chain, tethered together by disulfide bonds. The β-chain has multiple tyrosine phosphorylation sites, including three autophosphorylation sites at its activation loop. The overall structure of the IR is highly homologous to the IGF-I Receptor, except in their c-termini, where the two proteins diverge somewhat. The IR signals primarily by phosphorylating the Insulin Receptor Substrate (IRS) family of proteins, which creates docking sites for SH2-domain containing proteins. Insulin signaling is highly dependent on the PI3 Kinase pathway and Akt, which appear to mediate the functions of insulin.

Immunogen

Epitope: C-terminus
Recombinant protein corresponding to the C-terminus of human Insulin Receptor, beta subunit.

Application

Detect Insulin Receptor using this Anti-Insulin Receptor Antibody, beta subunit, clone CT-3 validated for use in WB, IH.
Research Category
Metabolism
Research Sub Category
Insulin/Energy Signaling

Biochem/physiol Actions

This antibody recognizes the C-terminus of human Insulin Receptor, beta subunit.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse brain tissue lysate
Evaluated by Western Blot in mouse brain tissue lysate.

Western Blot Analysis: 0.5 µg/ml of this antibody detected Insulin receptor beta subunit on 10 µg of mouse brain tissue lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
MABS6504053252397974