Skip to Content

MABS62

Anti-GREB1 Antibody, clone GREB1Ab

clone GREB1Ab, from mouse

Synonym(s):

growth regulation by estrogen in breast cancer 1, Gene regulated in breast cancer 1 protein, protein GREB1, gene regulated by estrogen in breast cancer protein

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
GREB1Ab, monoclonal
Application:
ICC, IHC, WB
Citations:
4
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

GREB1Ab, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable, immunohistochemistry: suitable, western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GREB1(9687)

General description

GREB1, also known as gene regulated in breast cancer protein 1 is shown to be involved in the proliferation of breast cancer cells, controlled by estrogen. GREB1 has the potential to be a clinical marker for response to endocrine therapy and a therapeutic target. GREB1 also may be the regulator for prostate cancer.
~ 216 kDa observed. Uncharacterized bands may be observed in some lysates.

Immunogen

Recombinant protein corresponding to human GREB1.

Application

Immunohistochemistry Analysis: 1:600 dilution from a previous lot detected GREB1 in prostate tissue.

Immunocytochemistry Analysis: A previous lot was used by an independent laboratory in IP. (Hnatyszyn, H., et al. 2009). Breast Cancer Res Treat
DOI 10.1007/s10549-009-0584-x.)
Research Category
Signaling

Apoptosis & Cancer
Research Sub Category
Hormones & Receptors
This Anti-GREB1 Antibody, clone GREB1Ab is validated for use in WB, IH, IC for the detection of GREB1.

Physical form

Format: Purified
Protein G
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
MCF-7 cell lysate
Evaluated by Western Blot in MCF-7 cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected GREB1 on 10 µg of MCF-7 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library