Skip to Content
MilliporeSigma
All Photos(1)

Documents

MABS492

Sigma-Aldrich

Anti-Lamin-B1 Antibody, clone 8F10.1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

antibody form

purified antibody

Quality Level

clone

8F10.1
monoclonal

purified by

using protein G

species reactivity

human, rat, mouse

concentration

(Please refer to lot specific datasheet.)

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG2bκ

UniProt accession no.

General description

GST-tagged recombinant protein corresponding to the Internal region of human Lamin-B1.

Application

Epitope: Internal region

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Lamin-B1 in 10 µg of HEK293 cell lysate.

Physical properties

~66 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service