Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MABS487

Sigma-Aldrich

Anti-Modified Citrulline Antibody, clone C4

clone C4, from human(Recombinant)

Synonym(s):

citrulline, modified citrulline

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

human (Recombinant)

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

C4, monoclonal

species reactivity (predicted by homology)

all

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG1λ

shipped in

wet ice

target post-translational modification

unmodified

General description

Citrulline is thought to be an unincorporated amino acid that plays a part in intermediary metabolism. It is produced in the small intestine by enterocytes. Citrulline concentration in plasma and serum is thought to be a reliable and noninvasive biomarker of small intestine function and mass during disease. Citrulline has also been shown to play an important role in protein homeostasis. Modified citrulline is a chemically modified form of citrulline that is more immunogenic.

Immunogen

Citrulline-containing peptide modified with 2,3-butanedione monoxime and antipyrine.

Application

Research Category
Signaling
Research Sub Category
Signaling Neuroscience
This Anti-Modified Citrulline Antibody is validated for use in Western blotting and ELISA for the detection of Modified Citrulline.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Modified Citrulline in 10 µg of treated HL-60 cell lysate, using Anti-Human secondary antibody.
ELISA Analysis: A representative lot detected Modified Citrulline in ELISA.

Quality

Evaluated by Western Blotting in treated cornified human cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Modified Citrulline in 10 µg of treated cornified human cell lysate, using Anti-Human secondary antibody.

Physical form

Format: Purified
Human recombinant monoclonal IgG1λ in buffer containing PBS with 0.05% sodium azide.
Protein A purified

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Association of citrullination with the progression of aortic stenosis.
Kim, et al.
Scientific Reports, 13, 8919-8919 (2023)
Issa O Yusuf et al.
Neurobiology of disease, 192, 106414-106414 (2024-01-23)
Alteration in protein citrullination (PC), a common posttranslational modification (PTM), contributes to pathogenesis in various inflammatory disorders. We previously reported that PC and protein arginine deiminase 2 (PAD2), the predominant enzyme isoform that catalyzes this PTM in the central nervous
Katerina D Samara et al.
Respiratory research, 18(1), 218-218 (2017-12-31)
Increased protein citrullination and peptidylarginine deiminases (PADIs), which catalyze the citrullination process, are central in Rheumatoid arthritis pathogenesis and probably involved in the initial steps towards autoimmunity. Approximately, 10% of RA patients develop clinically significantly ILD. A possible shared role
Seema Qamar et al.
Cell, 173(3), 720-734 (2018-04-21)
Reversible phase separation underpins the role of FUS in ribonucleoprotein granules and other membrane-free organelles and is, in part, driven by the intrinsically disordered low-complexity (LC) domain of FUS. Here, we report that cooperative cation-π interactions between tyrosines in the
Melissa M Ledet et al.
BMC cancer, 18(1), 412-412 (2018-04-14)
Mammary cancer is highly prevalent in dogs and cats and results in a poor prognosis due to critically lacking viable treatment options. Recent human and mouse studies have suggested that inhibiting peptidyl arginine deiminase enzymes (PAD) may be a novel

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service