Skip to Content
MilliporeSigma
All Photos(2)

Documents

MABS416

Sigma-Aldrich

Anti-ABCG5, clone 1B5E10 Antibody

clone 1B5E10, from mouse

Synonym(s):

ATP-binding cassette sub-family G member 5, Sterolin-1, ABCG5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

1B5E10, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
flow cytometry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ABCG5(64240)

General description

ATP-binding cassette sub-family G member 5, also known as Sterolin-1, and encoded by the gene name ABCG5, plays an important role in the selective transport of the dietary cholesterol in and out of the enterocytes and in the selective sterol excretion by the liver into bile. ABCG5 is localized too the membrane and is expressed in the liver, lower levels in the small intestine and colon. Mutations associated with ABCG5 is Sitosterolemia, which is a rare autosomal recessive disorder characterized by increased intestinal absorption of all sterols including cholesterol, plant and shellfish sterols, and decreased biliary excretion of dietary sterols into bile.

Immunogen

Purified recombinant fragment of human ABCG5 expressed in E. Coli

Application

Anti-ABCG5, clone 1B5E10 Antibody is an antibody against ABCG5 for use in western blotting, IHC, flow cytometry & ELISA.
Immunohistochemistry Analysis: A 1:200-1,000 dilution from a representative lot detected ABCG5 in cervical and rectum cancer tissues.

Flow Cytometry Analysis: A 1:200-400 dilution from a representative lot detected ABCG5 in A549 cells.

ELISA: A representative lot detected control Antigen (100 ng) and Antigen (10 ng, 50 ng, 100 ng) in ELISA (Direct).

Quality

Evaluated by Western Blotting in HL7702, Raji and Jurkat cell lysates.

Western Blotting Analysis: A 1:500-2,000 dilution of this antibody detected ABCG5 in HL7702, Raji and Jurkat cell lysates.

Target description

~70 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Linkage

Replaces: MABN1076

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service