Skip to Content
MilliporeSigma
All Photos(1)

Documents

MABS1896

Sigma-Aldrich

Anti-RNF123 Antibody, clone 2C10.1

clone 2C10.1, from mouse

Synonym(s):

E3 ubiquitin-protein ligase RNF123, EC 2.3.2.27, Kip1 ubiquitination-promoting complex protein 1, RING finger protein 123, RING-type E3 ubiquitin transferase RNF123

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2C10.1, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... RNF123(63891)

General description

E3 ubiquitin-protein ligase RNF123 (UniProt: Q5XPI4; also known as EC 2.3.2.27, Kip1 ubiquitination-promoting complex protein 1, RING finger protein 123, RING-type E3 ubiquitin transferase RNF123) is encoded by the RNF123 (also known as KPC1, PF1477) gene (Gene ID: 63891) in human.RNF123 serves as a catalytic subunit of the KPC complex that acts as E3 ubiquitin-protein ligase. It is required for poly-ubiquitination and proteasome-mediated degradation of CDKN1B/p27 during the cell cycle G1 phase of the cell cycle. It contains a ring-type zinc finger domain (aa 1254-1292). Two isoforms of RNF123 have been reported that are produced by alternative splicing. RNF123 is also reported to serve as a negative regulator of RIG-I and MDA5. Overexpression of RNF123 is shown to inhibit IFN-beta production triggered by Sendai virus (SeV) and encephalomyocarditis picornavirus (EMCV) and knockdown of endogenous RNF123 can potentiate IFN-beta production triggered by SeV and EMCV. RNF123 is shown to interact with the N-terminal CARD domains of RIG-I/MDA5 and compete with the downstream adaptor VISA/MAVS/IPS-1/Cardif for RIG-I/MDA5 CARD binding. (Ref.: Wang, S., et al. (2016). EMBO Rep. 17(8):1155-68).

Specificity

Clone 2C10.1 specifically detects RING-type E3 ubiquitin transferase RNF123 in human cells. It targets an epitope within 92 amino acids from the N-terminal region.

Immunogen

GST/His-tagged recombinant fragment corresponding to 92 amino acids from the N-terminal region of human RING-type E3 ubiquitin transferase RNF123.

Application

Anti-RNF123, clone 2C10.1, Cat. No. MABS1896, is a mouse monoclonal antibody that detects RING-type E3 ubiquitin transferase RNF123 and has been tested for use in Western Blotting.

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected RING-type E3 ubiquitin transferase RNF123 in HeLa cell lysate.

Target description

~150 kDa observed; 148.52 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service