Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABS1680

Sigma-Aldrich

Anti-PTEN-alpha Antibody, clone 3A4.1

clone 3A4.1, from mouse

Synonym(s):

Phosphatidylinositol 3, 4, 5-trisphosphate 3-phosphatase and dual-specificity protein phosphatase PTEN-alpha, Mitochondrial PTENalpha, Phosphatase and tensin-like protein-alpha, Mutated in multiple advanced cancers 1-alpha, Mitochondrial phosphatase and

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

3A4.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

General description

Phosphatidylinositol 3,4,5-trisphosphate 3-phosphatase and dual-specificity protein phosphatase PTEN (EC 3.1.3.16, EC 3.1.3.48, EC 3.1.3.67; UniProt P60484; also known as Mitochondrial PTENalpha, Phosphatase and tensin-like protein, Mutated in multiple advanced cancers 1, Mitochondrial phosphatase and tensin protein alpha, phosphatase and tensin homolog deleted on chromosome 10) is encoded by the PTEN (also known as 10q23del, BRRS, BZS, CWS1, GLM2, MMAC1, MHAM, TEP1) gene (Gene ID 5728) in human. PTEN is a tumor suppressor that catalyzes the removal of phosphate from phosphoserine, phosphothreonine, and phosphotyrosine residues on substrate proteins, as well as the phosphate in the D3 position of the inositol ring from phosphatidylinositol 3,4,5-trisphosphate, phosphatidylinositol 3,4-diphosphate, phosphatidylinositol 3-phosphate and inositol 1,3,4,5-tetrakisphosphate. PTEN plays a major role in controlling cell growth and proliferation, and PTEN gene mutains are found in a large number of cancers at high frequency. PTEN activity, stability, localization, and conformation are controlled through complex regulatory mechanisms, including the reversible phosphorylations of several Ser/Thr (S/T) residues in its unfolded C-terminal tail region (PTEN-Ctail, residues 351 403). Alternative translation start site 519 base pairs upstream of the ATG initiation sequence results in the translational variant of PTEN-alpha (PTEN-Long), including additional 173 N-terminal amino acids to the canonical 403-a.a. PTEN sequence. PTEN-Long is a membrane-permeable phosphatase secreted by PTEN-Long producing cells and can enter other cells as an exogenous PI3K signaling-inhibitory agent. PTEN-Long is reported to induce tumor cell death in vitro and in vivo.

Specificity

Clone 3A4.1 targets an N-terminal region specific to the spliced isoform alpha (PTEN-Long; P60484-2) and not present in spliced isoform 1 (a.k.a. 55 kDa isoform) or isoform 3.

Immunogen

GST-tagged recombinant human PTEN-alpha N-terminal fragment.

Application

Detect PTEN-alpha using this mouse monoclonal Anti-PTEN-alpha, clone 3A4.1 Antibody, Cat. No. MABS1680, validated for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected PTEN-alpha in human cerebellum, pancreas, and small intestine tissue sections.

Quality

Evaluated by Western Blotting in MCF-7 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected PTEN-alpha in 10 µg of MCF-7 cell lysate.

Target description

~78 kDa observed. 64.88 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Purified mouse monoclonal antibody IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4) 150 mM NaCl with 0.05% sodium azide

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magdalena C E Jochner et al.
Scientific reports, 9(1), 3183-3183 (2019-03-01)
Phosphatase and tensin homolog (PTEN) signalling might influence neuronal survival after brain ischemia. However, the influence of the less studied longer variant termed PTEN-L (or PTENα) has not been studied to date. Therefore, we examined the translational variant PTEN-L in
Liming Wang et al.
Cell research, 28(8), 787-802 (2018-06-24)
Mitophagy is an important type of selective autophagy for specific elimination of damaged mitochondria. PTEN-induced putative kinase protein 1 (PINK1)-catalyzed phosphorylation of ubiquitin (Ub) plays a critical role in the onset of PINK1-Parkin-mediated mitophagy. Phosphatase and tensin homolog (PTEN)-long (PTEN-L)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service