Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABS1310

Sigma-Aldrich

Anti-Apolipoprotein A-I Antibody, clone Wt20-7

clone Wt20-7, from mouse

Synonym(s):

Apo-AI, ApoA-I, Apolipoprotein A1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

Wt20-7, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

ELISA: suitable
dot blot: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... APOA1(335)

General description

Apolipoprotein A-I (UniProt: P02647; also known as Apo-AI, ApoA-I, Apolipoprotein A1) is encoded by the APOA1 gene (Gene ID: 335) in human. Apo A-1 is a major protein of plasma high density lipoprotein (HDL) and is also found in chylomicrons. It plays a major role in transporting cholesterol from peripheral tissues to the liver. It is synthesized mainly in the liver with a signal peptide (aa 1-18), which is proteolytically cleaved to generate the mature protein. Apo A-1 also serves as a co-factor for lecithin cholesterolacyltransferase (LCAT) that is involved in the formation of cholesteryl esters. Elevated levels of oxidized Apo A-1 (on Methionine 110 and 136) have been linked to increased risk for coronary artery disease. Mutations in APOA1 gene have also been linked to HDL deficiencies that lead to premature coronary artery disease and a reduction in cellular cholesterol efflux. Mutations in APOA1 gene have also shown to result in amyloidosis with extensive visceral amyloid deposit and in the pathogenesis of amyloid polyneuropathy-nephropathy Iowa type, also known as familial amyloid polyneuropathy type III.

Specificity

Clone Wt20-7 specifically detects human Apolipoprotein A-I.

Immunogen

GST-tagged full-length human recombinant Apolipoprotein A-I

Application

Anti-Apolipoprotein A-I, clone Wt20-7, Cat. No. MABS1310, is a mouse monoclonal antibody that detects human Apolipoprotein A-I and has been tested for use in Dot Blot, ELISA, Immunocytochemistry, and Western Blotting.
ELISA Analysis: A representative lot detected Apolipoprotein A-I in ELISA applications (Kimura, H., et. al. (2017). Sci Rep. 7(1):2988).

Western Blotting Analysis: A representative lot detected Apolipoprotein A-I in Western Blotting applications (Kuwabara, K., et. al. (2015). J Biol Chem. 290(40):24210-21).

Immunocytochemistry Analysis: A representative lot detected Apolipoprotein A-I in Immunocytochemistry applications (Kameyama, H., et. al. (2016). Sci Rep. 6:30391; Kuwabara, K., et. al. (2015). J Biol Chem. 290(40):24210-21).

Dot Blot Analysis: A representative lot detected Apolipoprotein A-I in Dot Blot applications (Kimura, H., et. al. (2017). Sci Rep. 7(1):2988; Kameyama, H., et. al. (2016). Sci Rep. 6:30391; Kuwabara, K., et. al. (2015). J Biol Chem. 290(40):24210-21).
Research Category
Signaling

Quality

Evaluated by Western Blotting in human fetal liver tissue lysate.

Western Blotting Analysis: A 1:2,000 dilution of this antibody detected Apolipoprotein A-I in human fetal liver tissue lysate.

Target description

~28 kDa observed; 30.78 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service