Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MABS1208

Sigma-Aldrich

Anti-LDL Receptor Antibody, clone 2H7.1

clone 2H7.1, from mouse

Synonym(s):

Low-density lipoprotein receptor, LDL Receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2H7.1, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LDLR(3949)

General description

Low-density lipoprotein receptor (UniProt P01130; also known as LDL receptor) is encoded by the LDLR (also known as FH, FHC) gene (Gene ID 3949) in human. Low-density lipoprotein (LDL) receptor recognizes the apoprotein B100 embedded in the outer phospholipid layer of cholesterol-rich LDL particles and mediates their remval by endocytosis via clathrin-coated pits. LDL receptor also recognizes the apoE protein found in chylomicron remnants and VLDL remnants (IDL). LDLR gene mutations are known causes of familial hypercholesterolaemia (FH). LDL receptor is initially produced with a signal peptide sequence (a.a. 1-21), the removal of which yields the mature single transmembrane (a.a. 789-810) protein with a C-terminal cytoplasmic tail (a.a. 811-860) and a large N-terminal extracellular region (a.a. 22-788) that contains seven class A (a.a. 25-313) and six class B (a.a. 397-658) LDLR repeats, as well as three EGF-like domains (a.a. 314-353, 354-393, 663-712). LDL-A repeats mediate LDL binding, repeats 2-7 are required for ApoB binding, while only repeat 5 is required for ApoE binding. The class B (LDL-B) repeats region adopts a beta-propeller conformation and is believed to be responsible for the pH-dependent release of LDL in the endosome. The cytoplasmic tail contains a signal sequence important for localizing the receptors to clathrin-coated pits and for triggering receptor-mediated endocytosis upon LDL binding.

Specificity

Expected to react with spliced isoforms 1 and 5 reported by UniProt P01130). Reactivity toward spliced isoforms 2, 3, 4, and 6 has not been determined.

Immunogen

Epitope: Extracellular class A domains 1-3.
GST-tagged recombinant human LDL receptor extracellular fragment corresponding to class A domains 1-3.

Application

Anti-LDL Receptor Antibody, clone 2H7.1 is an antibody against LDL Receptor for use in Western Blotting, Immunohistochemistry (Paraffin), Flow Cytometry.
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected LDL receptor immunoreactivity in human adrenal gland tissue.
Flow Cytometry Analysis: 1.0 µg of this antibody from a representative lot detected LDL receptor expression in one million HepG2 cells.
Research Category
Signaling
Research Sub Category
Lipid Metabolism & Weight Regulation

Quality

Evaluated by Western Blotting in human adrenal gland tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected LDL receptor in 10 µg of human adrenal gland tissue lysate.

Target description

~150 kDa observed. Target band size appears larger than the calculated molecular weights of 95.38 kDa (isoform 1), 75.85 kDa (isoform 2), 76.86 kDa (isoform 3), 90.94 kDa (isoform 4), 95.09 kDa (isoform 5), and 82.25 kDa (isoform 6) due to glycosylation. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Youxi Zhang et al.
Bioorganic & medicinal chemistry, 27(11), 2187-2191 (2019-04-22)
Targeted drug delivery platforms can increase the concentration of drugs in specific cell populations, reduce adverse effects, and hence improve the therapeutic effect of drugs. Herein, we designed two conjugates by installing the targeting ligand GalNAc (N-acetylgalactosamine) onto atorvastatin (AT).
Xinfu Zhang et al.
Bioconjugate chemistry, 28(8), 2109-2113 (2017-07-27)
Targeting ligands facilitate cell specific drug delivery and improve pharmaceutical properties. Herein, we designed two ligand drug conjugates by conjugating GlcNAc (N-acetylglucosamine) with atorvastatin. These two conjugates, termed G-AT and G-K-AT, exhibited enhanced water solubility and cellular uptake. Moreover, both
Shyam Sharan Shrestha et al.
Molecules (Basel, Switzerland), 25(7) (2020-04-03)
Girardinia diversifolia, also known as Himalayan nettle, is a perennial herb used in Nepal to make fiber as well as in traditional medicine for the treatment of several diseases. To date, phytochemical studies and biological assays on this plant are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service