Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MABN839

Sigma-Aldrich

Anti-TrpV6 Antibody, clone 4A5.1

culture supernatant, clone 4A5.1, from mouse

Synonym(s):

Transient receptor potential cation channel subfamily V member 6, Alu-binding protein with zinc finger domain, Calcium transport protein 1, Calcium transporter-like protein, CaT-L, CaT-like, CaT1, ECaC2, Epithelial apical membrane calcium transporter/cha

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

4A5.1, monoclonal

species reactivity

mouse, human, rat

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

General description

Transient receptor potential cation channel subfamily V member 6 (UniProt Q9H1D0; also known as Alu-binding protein with zinc finger domain, Calcium transport protein 1, Calcium transporter-like protein, CaT-L, CaT-like, CaT1, ECaC2, Epithelial apical membrane calcium transporter/channel CaT1, Epithelial calcium channel 2, TrpV6) is encoded by the TRPV6 (also known as ABP/ZF, CAT1, CATL, ECAC2, ZFAB) gene (Gene ID 55503) in human. TRPV6 belongs to the transient receptor potential (TRP) family of membrane proteins. The human TRPV6 is an 765-amino acid, six-transmembrane protein with both N- and C-termini exposed intracellularly. TRPV6 contains a pore-forming loop domain located between the fifth and sixth transmembrane region. Among all the TRPV family members, TRPV5 and TRPV6 are the two highly Ca2+-selective channels that play pivotal roles in the Ca2+-reabsorption in the kidney and intestine, respectively. TRPV6 is found in the apical brush-border membrane of the intestinal enterocytes where it regulates calcium entry into the cells. TRPV6 expression is also reported in human placenta, pancreas, and prostate gland. Vitamin D treatment of human colon cancer Caco-2 cells and duodenal explants results in upregulated TRPV6 mRNA levels.

Specificity

Expected to react with both spliced isoforms of human TrpV6 reported by UniProt (Q9H1D0).

Immunogen

Epitope: C-terminal region.
GST-tagged recombinant human TrpV6 C-terminal fragment.

Application

Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience
Western Blotting Analysis: A 1:500 dilution from a representative lot detected TrpV6 in 10 µg of LNCaP prostate adenocarcinoma and K562 erythroleukemia cell lysates.
Immunohistochemistry Analysis: A 1:50-1:250 dilution from a representative lot detected TrpV6 in human pancreas, human small intestine, rat colon tissue, and mouse brain tissues.

Quality

Evaluated by Western Blotting in Y79 retinoblastoma cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected TrpV6 in 10 µg of Y79 retinoblastoma cell lysate.

Target description

~70 kDa observed.

Physical form

In culture supernatant without preservatives.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service