Skip to Content
MilliporeSigma
All Photos(4)

Documents

MABN695

Sigma-Aldrich

Anti-CRTC2 Antibody, clone 5B10

ascites fluid, clone 5B10, from mouse

Synonym(s):

TORC2, TORC-2, CRTC2, CREB-regulated transcription coactivator 2, Transducer of regulated cAMP response element-binding protein 2, Transducer of CREB protein 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

5B10, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CRTC2(200186)

General description

CREB-regulated transcription coactivator 2 (CRTC2) is a transcriptional coactivator for CREB1 which is a cAMP (CRE) site responder. CRTC2 is also known as TORC-2 (transducer of regulated cAMP response element-binding protein 2) and CRTC2 regulates gluconeogenesis as well as steroidogenic processes, and is also a coactivator for mitochondrial biogenesis in muscle cells. Localization is cytoplasmic but relocates to the nucleus in response to cAMP and glucagon levels. CRTC2 is highly expressed in thymus, spleen, muscle, and lung; it is also expressed in various cancers as well as insulinomas. Low levels of expression are found in the brain, colon, heart and kidney, with the lowest levels being in the liver and pancreas. Activity is regulated via phosphorylation. CRTC2 is inactive when phosphorylated (meaning that is does not bind CREB and assist in gene transcription) and active when dephosphorylated. EMD-Millipore’s Anti-CRTC2 mouse monoclonal, clone 5B10, has been tested in western blots against human HEK292 and HeLa cell extracts, in immunohistochemistry in paraffin embedded human ovary and lung cancer tissue, and in immunofluorescent immunocytochemistry and flow cytometry on HeLa cells in culture.

Immunogen

Purified protein fragment of human CRTC2 expressed in E. coli.

Application

Anti-CRTC2 Antibody, clone 5B10 is a highly specific mouse monoclonal antibody, that targets CREB-regulated transcription coactivator 2 & has been tested in western blotting, ICC, IHC & Flow Cytometry.
Immunocytochemistry Analysis: A 1:200-1,000 dilution from a representative lot detected CRTC2 in HeLa cells.

Immunohistochemistry Analysis: A 1:200-1,000 dilution from a representative lot detected CRTC2 in human lung cancer and ovarian tumor tissues.

Flow Cytometry Analysis: A 1:200-400 dilution from a representative lot detected CRTC2 in HeLa cells.

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected CRTC2 in 10 µg of HeLa cell lysate.

Target description

~76 kDa observed.

Physical form

Mouse monoclonal IgG2b ascitic fluid containing up to 0.1% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
HeLa cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service