Skip to Content
MilliporeSigma
All Photos(2)

Documents

MABN666

Sigma-Aldrich

Anti-TRIP8b Antibody, clone N212A/34

clone N212A/34, from mouse

Synonym(s):

PEX5-related protein, PEX5-like protein, Peroxin-5-related protein, TPR-containing Rab8b-interacting protein, Tetratricopeptide repeat-containing Rab8b-interacting protein, Pex5Rp, TRIP8b

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

N212A/34, monoclonal

species reactivity

human, rat, mouse

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

Related Categories

General description

Tetratricopeptide repeat-containing Rab8b-interacting protein (TRIP8b) is also called PEX5-related protein, PEX5-like protein and Peroxin-5-related protein (Pex5Rp). TRIP8b a hyperpolarization-activated cyclic nucleotide-gated (HCN) channel subunit that is involved in HCN cynclic nucleotide dependence and expression at the cell surface. TRIP8b is expressed specifically in the brain.

Specificity

This antibody demonstrates specificity for TRIP8b containing exon 1b. It does not cross react with TRIP8b containing exons 1a, 2, or 4.

Immunogen

Recombinant protein corresponding to rat TRIP8b.

Application

Anti-TRIP8b Antibody, clone N212A/34 is a highly specific mouse monoclonal antibody & that targets Tetratricopeptide repeat-containing Rab8b-interacting protein & has been tested in western blotting.
Research Category
Neuroscience
Research Sub Category
Synapse & Synaptic Biology
Western Blotting Analysis: 0.5 μg/mL of this antibody detected TRIP8b in 10 μg of human brain tissue lysate.

Western Blotting Analysis: A representative lot detected TRIP8b in COS cell lysates transiently transfected with GFP tagged TRIP8b (exon1b) and not in COS cell lysates transiently transfected with TRIP8b (exon1a, 2, or 4) or with untagged Kv2.1 plasmid (Prof. J. Trimmer, University of California, Davis.).

Quality

Evaluated by Western Blotting in rat brain tissue lysate.

Western Blotting Analyis: 0.5 μg/mL of this antibody detected TRIP8b in 10 μg of rat brain tissue lysate.

Target description

~70 kDa observed. Multiple isoforms may be observed due to alternative splicing (Lewis, A. S., et al. (2009). J Neurosci. 29(19):6250-6265.). Uncharacterized band(s) may be observed in some cell lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Rat brain tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service