Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MABN491

Sigma-Aldrich

Anti-Brevican Antibody, clone N294A/6

clone N294A/6, from mouse

Synonym(s):

Brevican core protein, Brain-enriched hyaluronan-binding protein, BEHAB, Brevican

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

N294A/6, monoclonal

species reactivity

rat, human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BCAN(63827)

General description

Brevican core protein (BCAN), also known as Brain-enriched hyaluronan-binding protein (BEHAB) or Chondroitin sulfate proteoglycan 7 (CSPG7), may play a role in the terminally differentiating and the adult nervous system during postnatal development. It may stabilize interactions between hyaluronan (HA) and brain proteoglycans. Two isoforms exist, with isoform 1 (~99 kDa) being highly expressed from birth through 8 years of age and down-regulated by 20 years of age to low levels that are maintained in the normal adult cortex; and isoform 2 (~72 kDa) being expressed at uniformly low levels throughout development.

Immunogen

Recombinant protein corresponding to rat Brevican.

Application

Anti-Brevican, clone N294A/6 is an antibody targeting the Brevican protein, validated for use in WB & IHC.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience
Western Blot Analysis: 2 µg/mL from a representative lot detected Brevican in 10 µg of human brain tissue lysate.

Immunohistochemistry Analysis: A 1:500 dilution from a representative lot detected Brevican in rat midbrain, rat thalamus, and rat cerebellum tissues.

Western Blot Analysis: A representative lot detected Brevican in rat brain tissue lysate and in WT, but not in Brevican knockout mouse brain tissue lysate.

Quality

Evaluated by Western Blot in rat brain tissue lysate.

Western Blot Analysis: 2 µg/mL of this antibody detected Brevican in 10 µg of rat brain tissue lysate.

Target description

~140 kDa observed. The calculated molecular weight of this protein is 96 kDa A second isoform is known to exist at 69 kDa This protein may be observed at a higher molecular weight due to glycosylation. A proteolyticaly cleaved fragment of Brevican may be observed at ~50 kDa in some tissue lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Rat brain tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ditte S Jonesco et al.
PloS one, 15(6), e0234632-e0234632 (2020-06-20)
Evidence indicate that the brain-specific protein, brevican, is proteolytically cleaved during neurodegeneration, hence positioning fragments of brevican as potential blood biomarkers of neurodegenerative diseases, such as dementia. We aimed to develop two assays capable of detecting the brevican N-terminal (N-Brev)
Moon Hang Kim et al.
Tissue engineering and regenerative medicine, 18(1), 165-178 (2020-09-18)
Chondroitin sulfate glycosaminoglycans (CS-GAGs) are the primary inhibitory GAGs for neuronal growth after central nervous system (CNS) injury. However, the inhibitory or permissive activity of CS-GAG subtypes is controversial and depends on the physiological needs of CNS tissues. In this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service