Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABN40

Sigma-Aldrich

Anti-LRRK2 Antibody, clone N138/6

clone N138/6, from mouse

Synonym(s):

leucine-rich repeat kinase 2, DARDARIN, augmented in rheumatoid arthritis 17, Parkinson disease (autosomal dominant) 8, Dardarin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

N138/6, monoclonal

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LRRK2(120892)

General description

Leucine-rich repeat kinase 2 (LRRK2) is a 2,527 amino acid protein encoded by the PARK8 gene and belonging to the ROCO family of proteins. This protein family is characterized as having a unique interaction between the Roc domain and the intrinsic GTPase activity along with a COR domain. LRKK2 consists of 5 different domains; a tyrosine kinase-like MAPKKK-related domain, WD40 propeller motif repeats, a Roc (Ras-like small GTPase family) domain, a COR (C-terminal of Roc) domain, and an LRRK. LRRK2 is mainly localized to the cytoplasm and expression within tissue is most highly observed within the brain. It is found in lower levels within the liver, heart, lung, skeletal muscles, placenta, pancreas, and kidney. The exact function of LRRK2 is unknown; however mutations to any of the five domains are the most commonly known causes of Parkinson’s disease, both familial and sporadic. Further study of this protein may lead to a better understanding of the disease’s pathogenesis, as well as potential therapeutic markers.

Specificity

This antibody recognizes LRRK2.

Immunogen

Recombinant protein corresponding to human LRRK2.

Application

Anti-LRRK2 Antibody, clone N138/6 detects level of LRRK2 & has been published & validated for use in IH & WB.
Western Blot Analysis: A previous lot was used by an independent laboratory in WB. (UC Davis/NIH NeuroMab Facility, www.neuromab.org, neuromab@ucdavis.edu)

Quality

Evaluated by Immunohistochemistry in human placenta and Parkinson′s diseased vascular brain tissue.

Immunohistochemistry Analysis: 1:300 dilution of this antibody detected LRRK2 on 10 µg of human placenta and Parkinson′s diseased vascular brain tissue.

Target description

240 kDa observed. 286 kDa calculated. (Glasson, B., et al. (2006). Ann. Neurol. 59(2):315-322.)

Physical form

Format: Purified

Analysis Note

Control
Human placenta and Parkinson′s diseased vascular brain tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iakov N Rudenko et al.
The Biochemical journal, 474(9), 1547-1558 (2017-03-23)
Autosomal dominant mutations in leucine-rich repeat kinase 2 (LRRK2) are associated with Parkinson's disease (PD). Most pathogenic LRRK2 mutations result in amino acid substitutions in the central ROC (Ras of complex proteins)-C-terminus of ROC-kinase triple domain and affect enzymatic functions
Ruth Chia et al.
Nature communications, 5, 5827-5827 (2014-12-17)
LRRK2, a gene relevant to Parkinson's disease, encodes a scaffolding protein with both GTPase and kinase activities. LRRK2 protein is itself phosphorylated and therefore is subject to regulation by cell signalling; however, the kinase(s) responsible for this event have not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service