Skip to Content
MilliporeSigma
All Photos(2)

Documents

MABN2475

Sigma-Aldrich

Anti-NMDAR2B Antibody, clone 11D1

clone 11D1, from mouse

Synonym(s):

Glutamate receptor ionotropic NMDA 2B, GluN2B, Glutamate [NMDA] receptor subunit epsilon-2, N-methyl D-aspartate receptor subtype 2B, NR2B, N-methyl-D-aspartate receptor subunit 3, NR3, hNR3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

11D1, monoclonal

species reactivity

mouse, human, rat

packaging

antibody small pack of 25 μg

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... GRIN2B(2904)
mouse ... Grin2B(14812)
rat ... Grin2B(24410)

General description

Glutamate receptor ionotropic, NMDA2B (UniProt: Q13224; also known GluN2B, Glutamate [NMDA] receptor subunit epsilon-2, N-methyl D-aspartate receptor subtype 2B, NMDAR2B, NR2B, N-methyl-D-aspartate receptor subunit 3, NR3, hNR3) is encoded by the GRIN2B (also known as NMDAR2B) gene (Gene ID: 2904) in human. NMDA receptor complexes are ligand-gated ion channels with high calcium permeability and voltage-dependent sensitivity to magnesium. They are involved in long-term potentiation and neuronal plasticity and regulate the number of nerve cells during development. However, over-stimulation NMDA receptors can lead to neuronal degeneration. NMDAR2B is a heterotetrameric, multi-pass membrane protein that is synthesized with a signal peptide (aa 1-26), which is subsequently cleaved off in the mature form. The heterotetrameric channel is composed of two zeta subunits and two epsilon subunits, but can also form channels that contain at least one zeta subunit and at least one epsilon subunit. Activation of NMDA 2B channel requires binding of glutamate to the epsilon subunit, glycine binding to the zeta subunit, plus membrane depolarization to eliminate channel inhibition by magnesium ions. NMDA 2B is reported to be phosphorylated at Ser1303 by DAPK1 that enhances its channel activity allowing injurious Ca2+ influx that results in neuronal death. Mutations in NMDAR2B have been linked to early infantile epileptic encephalopathy that is characterized by refractory seizures and neurodevelopmental impairment. Some mutations are also known to cause mental retardation with or without seizures.

Specificity

Clone 11D1 is a mouse monoclonal antibody that specifically detects NMDAR2B.

Immunogen

Full length purified human Glutamate receptor ionotropic, NMDA 2B (NMDAR2B).

Application

Anti-NMDAR2B, clone 11D1, Cat. No. MABN2475, is a highly specific mouse monoclonal antibody that targets Glutamate receptor ionotropic, NMDA 2B and has been tested for use in Western Blotting.
Research Category
Neuroscience
Western Blotting Analysis: A representative lot detected NMDAR2B in Western Blotting applications (Lu, W., et. al. (2017). Science. 355(6331)).

Western Blotting Analysis: 4 µg/mL from a representative lot detected NMDAR2B in mouse brain tissue lysate.

Quality

Evaluated by Western Blotting in rat brain tissue lysate.

Western Blotting Analysis: 4 µg/mL of this antibody detected NMDAR2B in rat brain tissue lysate.

Target description

166 kDa observed; 166.37 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service