Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABN1581

Sigma-Aldrich

Anti-CLCA4 Antibody, clone 1C5.1

clone 1C5.1, from mouse

Synonym(s):

Calcium-activated chloride channel regulator 4, CaCC-2, Calcium-activated chloride channel family member 4, Calcium-activated chloride channel protein 2, hCaCC-2, hCLCA4, Calcium-activated chloride channel regulator 4, 110 kDa form

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1C5.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... CLCA4(22802)

General description

Calcium-activated chloride channel regulator 4 (UniProt Q14CN2; also known as CaCC-2, Calcium-activated chloride channel family member 4, Calcium-activated chloride channel protein 2, hCaCC-2, hCLCA4) is encoded by the CLCA4 (also known as CaCC2) gene (ORF UNQ562/PRO1124; Gene ID 22802) in human. CLCA4 (CaCC2) belongs to the calcium-activated chloride channel regulator (previously known as chloride channel calcium activated or CLCA) family of self-cleaving, secreted metalloproteases that activate calcium-dependent chloride currents (ICaCC) in mammalian cells. Originally thought to be calcium-activated chloride channels (CaCCs) themselves, CLCA proteins were later identified as secreted CaCCs modulators. Among the four CLCA genes identified in human genome, only three (CLCA1/CACC1, CLCA2/CACC3, CLCA4/CACC2) encode functional Cl channel accessory proteins. CLCA4 is predominantly expressed in colon along with CLCA1, and both are reportedly downregulated in colorectal cancer cells. CLCA4 is also reported to be downregulated in breast cancer cells and exogenous CLCA4 overexpression inhibits breast cancer cell proliferation, while CLCA4 knockdown induces epithelial-to-mesenchymal transition (EMT) in mammary epithelial cells. CLCA4 is produced with a signal peptide sequence (a.a. 1-21) that is removed posttranslationally. Similar to other CLCA members, CLCA4 is further processed proteolytically by its metalloprotease domain (a.a. 45-119) to yield a 110 kDa soluble form and a 30 kDa membrane-bound form. The soluble fragment of all CLCA members also contains a von Willebrand factor type A (VWA) domain that in the case of CLCA2 is shown to engage interaction with beta4-integrin.

Specificity

Clone 1C5.1 recognizes a metalloprotease domain epitope that is not present in the 30 kDa membrane-bound form or the spliced isoforms 2 reported by UniProt (Q14CN2).

Immunogen

Epitope: Metalloprotease domain.
GST-tagged recombinant human CLCA4 metalloprotease domain fragment (~40 kDa).

Application

Anti-CLCA4 Antibody, clone 1C5.1 is an antibody against CLCA4 for use in Western Blotting, Immunohistochemistry (Paraffin).
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected CLCA4 in human cerebellum, colon, kidney and testis tissue sections.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters

Quality

Evaluated by Western Blotting of human CLCA4 GST fusion protein.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected recombinant human CLCA4 metalloprotease domain fragment GST fusion protein.

Target description

101.3 kDa (pro-form) and 98.89 kDa (mature form) calculated. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service