Skip to Content
MilliporeSigma
All Photos(1)

Documents

MABF956

Sigma-Aldrich

Anti-Mouse CD36 Antibody, Clone 63

clone 63, from mouse

Synonym(s):

Glycoprotein IIIb, GPIIIB, PAS IV, PAS-4, Platelet glycoprotein IV, GPIV, CD antigen CD36

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

63, monoclonal

species reactivity

mouse, rat

technique(s)

flow cytometry: suitable
inhibition assay: suitable

isotype

IgAκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

mouse ... Cd36 (12491)

General description

Platelet glycoprotein 4 (UniProt: Q08857; also known as Glycoprotein IIIb, GPIIIB, PAS IV, PAS-4, Platelet glycoprotein IV, GPIV, CD antigen: CD36) is encoded by the cd36 gene (Gene ID: 12491) in murine species. CD36 is a multifunctional glycoprotein that acts as receptor for a broad range of ligands, including oxLDL. CD36 is shown to play a role in scavenging oxLDL may also mediate effects of oxLDL on monocytes and platelets in atherosclerotic lesions. Upon ligand-binding, it is internalized through dynamin-dependent endocytosis. CD36 is highly expressed in the intestine on the luminal surface of enterocytes. In small intestines expression levels follow a steep decreasing gradient from proximal to distal segments. It is expressed in a circadian manner in the circumvallate papillae with its levels being lower during the dark period. In the small intestine, plays a role in proximal absorption of dietary fatty acid and cholesterol for optimal chylomicron formation.CD36 null mice exhibit impaired chylomicron secretion, but no overall lipid absorption defect. CD36 can binds long-chain fatty acids and facilitates their transport into cells, thus participating in muscle lipid utilization, adipose energy storage, and gut fat absorption. In taste receptor cells, CD36 is shown to mediate the induction of an increase in intracellular calcium levels by long-chain fatty acids, leading to the activation of the gustatory neurons in the nucleus of the solitary tract. CD36 is also expressed in lung alveolar macrophages and dendritic macrophages. (Ref.: Nassir, F., et al. (2007). J. Biol. Chem. 282(27):19493-194501).

Specificity

Clone 63 specifically detects CD36 in RAW264.7 cells.

Immunogen

Full-length adenovirus vector injected into CD36 null mice.

Application

Detect CD36 by using this mouse monoclonal Anti-Mouse CD36 Antibody, Clone 63, Cat. No. MABF956. It is used in Flow Cytometry and Inhibition studies.
Flow Cytometry Analysis: A representative lot detected Mouse CD36 in Flow Cytometry applications (Corcoran, L., et. al. (2002). Int Immunol. 14(10):1099-104).

Inhibition Analysis: A representative lot detected Mouse CD36 in Inhibition applications (Patel, S.N., et. al. (2004). J Infect Dis. 189(2):204-13).

Quality

Evaluated by Flow Cytometry in RAW264.7 cells.

Flow Cytometry Analysis: 1 µg of this antibody detected CD36 in one million RAW264.7 cells.

Target description

52.70 kDa calculated.

Physical form

Format: Purified
Purified mouse monoclonal antibody IgA in PBS without preservatives.

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service