Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABF94

Sigma-Aldrich

Anti-BDCA-2 Antibody, clone 10E6.1

clone 10E6.1, from mouse

Synonym(s):

C-type lectin domain family 4 member C, Blood dendritic cell antigen 2, BDCA-2, C-type lectin superfamily member 7, Dendritic lectin, CD303

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

10E6.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CLEC4C(170482)

General description

Blood dendritic cell antigen 2 (BDCA-2;CD303) is a type II C-type lectin that is expressed on plasmacytoid dendritic cells (PDC), a subset of immature dendritic cells found in lymphoid and nonlymphoid tissues. BDCA-2 is critically involved in both adaptive and innate immune responses. It is involved in the recognition and internalization of pathogens, but inhibits TLR6 signaling and INF-1 production, in PDC cells, possibly through signaling of the BCR-like signalosome. PDC signaling has been linked to multiple inflammatory diseases such as granulomatous lymphadenitis.

Immunogen

GST-tagged recombinant protein corresponding to human BDCA-2.

Application

Detect BDCA-2 using this Anti-BDCA-2 Antibody, clone 10E6.1 validated for use in Western Blotting & IHC.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected BDCA-2 in human tonsil tissue.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Quality

Evaluated by Western Blot in recombinant BDCA-2 protein.

Western Blot Analysis: 1 µg/mL of this antibody detected BDCA-2 on 10 µg of recombinant BDCA-2 protein.

Target description

~33 kDa observed. Uncharacterized bands appear at ~25 and ~31 kDa in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Recombinant BDCA-2 protein

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service