Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABF536

Sigma-Aldrich

Anti-CD127 (IL-7Ra) Antibody (mouse), Biotin, clone A7R34

clone A7R34, 0.5 mg/mL, from rat

Synonym(s):

Interleukin-17 receptor A, IL-17 receptor A, IL-17RA, CD217

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rat

Quality Level

conjugate

biotin conjugate

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

A7R34, monoclonal

species reactivity

mouse

packaging

antibody small pack of 25 μg

concentration

0.5 mg/mL

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG2aκ

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Il7R(16197)

General description

The A7R34 antibody is specific for mouse CD127, a 60-90 kDa cell surface protein also known as the Interleukin-7 Receptor alpha chain, or IL-7R alpha. CD127 is typically expressed at the cell surface as a heterodimer with the common gamma chain (CD132). This complex acts as the functional receptor for IL-7, a cytokine important in T and B cell development, and in mature T cell homeostasis. A second cytokine known as Thymic Stromal Lymphopoietin (TSLP) also binds to a receptor complex of CD127 and the TSLPR chain to trigger activation of dendritic cells, and is involved in B cell development, allergy and autoimmunity.The A7R34 antibody may be used as a phenotypic marker for CD127 on immature B cells, on subsets of thymocytes which are double negative (CD4-CD8- ) or single positive (CD4+ or CD8+), and at low levels on mature, peripheral T cells. CD127 is a key marker, when used in combination with CD4 and CD25, to distinguish Treg and effector/memory Treg populations known as T(REM).
This rat monoclonal antibody Anti-CD127 (IL-7Ra) (mouse), Biotin, clone A7R34 validated for use in IP, Flow Cytometry & IHC for the detection of CD127 (IL-7Ra).

Quality

The monoclonal antibody was purified from tissue culture supernatant via affinity chromatography. The purified antibody was conjugated under optimal conditions, with unreacted dye removed from the preparation.

Flow Cytometry Analysis: 0.06 µg of this antibody detected CD127 (IL-7Ra) in one million C57Bl/6 splenocytes.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service