Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABF3136

Sigma-Aldrich

Anti-IL-11 Antibody, clone 13-6D-10G

Synonym(s):

Interleukin-11

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

13-6D-10G, monoclonal

mol wt

calculated mol wt 21.52 kDa
observed mol wt ~20 kDa

purified by

using protein G

species reactivity

mouse

packaging

antibody small pack of 100 μL

technique(s)

inhibition assay: suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

storage temp.

-10 to -25°C

target post-translational modification

unmodified

Gene Information

mouse ... Il11(16156)

General description

Interleukin-11 (UniProt: P47873 ; also known as IL-11) is encoded by the Il11 gene (Gene ID: 16156) in murine species. IL-11 is member of the IL-6 family of cytokines that is synthesized with a signal peptide (aa 1-21), which is subsequently cleaved off to produce mature secreted protein. It is involved in a variety of biological responses, including hematopoiesis, bone development, and carcinogenesis. Its actions are mediated via interaction with IL-11 receptor that is composed of IL-11R and gp130. Binding of IL-11 to its receptor induces the activation of JAK that phosphorylates STAT3, which then translocates to the nucleus and activates transcription of various genes. IL-11 is reported to promote the proliferation of hepatocytes in response to liver damage. In mice subjected to liver injury, IL-11 is mainly produced by hepatocytes in response to reactive oxygen species (ROS) released from dying hepatocytes. Secreted IL-11 then induces STAT3 phosphorylation in adjacent healthy hepatocytes resulting in their compensatory proliferation. It has been reported that IL-11 production in dying hepatocytes depended on ERK2 that phosphorylates Fra-1 and prevents its proteasomal degradation. Fra-1 is subsequently recruited to the Il11 promoter and activates transcription. Clone 13-6D-10G (13) displays neutralizing activity against mIL-11. It blocks mIL-11-induced, but not mIL-6-induced the proliferation of T1165 cells. Preincubation of murine IL-11 with this clone is shown to completely inhibit STAT3 phosphorylation. (Ref.: Deguchi, Y., et al. (2018). Biochem. Biophys. Res. Commun. 505(2); 453-459; Nishina, T., et al. (2012). Sci. Signal. 5(207); ra5).

Specificity

Clone 13-6D-10G is a rat monoclonal antibody that detects murine IL-11.

Immunogen

FLAG-tagged full-length recombinant murine IL-11 produced in HEK293T cells transfected with pcDNA3-mIl11.

Application

Quality Control Testing

Evaluated by Western Blotting in HEK293T cells transiently transfected with mIL-11.

Western Blotting Analysis (WB): A 1:250 dilution of this antibody detected IL-11 in HEK293T cells transiently transfected with mIL-11, but not in wild-type HEK293T cells.

Tested Applications

Inhibition Assay: A representative lot inhibited mIL-11 induced STAT3 phosphorylation and T1165 cell proliferation. (Deguchi, Y., et al. (2018). Biochem Biophys Res Commun. 505(2):453-459).

Western Blotting Analysis: A representative lot detected IL-11 in Western Blotting applications (Deguchi, Y., et al. (2018). Biochem Biophys Res Commun. 505(2):453-459).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Physical form

Purified rat monoclonal antibody IgG1 in PBS without azide.

Reconstitution

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service