Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABF235

Sigma-Aldrich

Anti-AIRE Antibody, clone Mab155

clone Mab155, 1 mg/mL, from mouse

Synonym(s):

Autoimmune regulator, Autoimmune polyendocrinopathy candidiasis ectodermal dystrophy protein, APECED protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

Mab155, monoclonal

species reactivity

human

concentration

1 mg/mL

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... AIRE(326)

General description

AIRE (Autoimmune Regulator) is a human gene that is expressed primarily in the thymus. It causes transcription of a wide selection of organ-specific genes, reducing the threat of autoimmunity occurring by allowing the elimination of autoreactive T cells via negative selection. Defects in this gene cause the rare autosomal-recessive systemic autoimmune disease termed autoimmune polyendo-crinopathy-candidiasis-ectodermal dystrophy (APECED). Disruption of AIRE results in the development of a range of autoimmune diseases, the most common clinical conditions in the syndrome are hypoparathyroidism, primary adrenocortical failure and chronic mucocutaneous candidiasis. Aire functions through initiating the transcription of a diverse set of self-antigens, such as insulin, in the thymus. This expression then allows maturing thymocytes to become tolerant towards peripheral organs, thereby suppressing autoimmune disease. Aire is atranscriptional regulator protein that binds to DNA as dimer and tetramer, but not as a monomer. It forms nuclear bodies and interacts with the transcriptional coactivator CBP (CREBBP). At least three splice variant mRNAs products have been described including one which results in a premature stop codon and a transcript predicted to be a candidate for nuclear-mediated decay (NMD).

Immunogen

His-tagged recombinant protein corresponding to human AIRE.

Application

Research Category
Inflammation & Immunology
Research Sub Category
Immunological Signaling
This Anti-AIRE Antibody, clone Mab155 is validated for use in Flow Cytometry and Western Blotting and Immunocytochemistry for the detection of AIRE.
Western Blotting Analysis: A representative lot detected AIRE in Cos1 transfected cell lysate (Rinderle, C., et al. (1999). Human Molecular Genetics. 8(2):277-290).
Immunocytochemistry Analysis: A representative lot detected AIRE in freshly isolated endogenous peripheral blood lymphocytes (Rinderle, C., et al. (1999). Human Molecular Genetics. 8(2):277-290).

Quality

Evaluated by Flow Cytometry in human PBMC cells.

Flow Cytometry Analysis: 0.5 µg of this antibody detected AIRE in human PBMC cells.

Target description

58 kDa calculated

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service