Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABF2032M

Sigma-Aldrich

Anti-CD31 (PECAM-1) Antibody, clone MBC 78.1

clone MBC 78.1, from mouse

Synonym(s):

Platelet endothelial cell adhesion molecule, PECAM-1, EndoCAM. GPIIA′, PECA1, CD31

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

MBC 78.1, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... PECAM1(5175)

General description

Platelet endothelial cell adhesion molecule (UniProt: P16284; also known as PECAM-1, EndoCAM. GPIIA′, PECA1, CD31) is encoded by the PECAM1 gene (Gene ID: 5175) in human. PECAM-1 is a single-pass type I membrane protein that is synthesized with a signal peptide (aa 1-27) that is subsequently cleaved to generate the mature protein with a large extracellular region (aa 28-601) that contains six Ig-like C2-type domains, followed by a transmembrane segment (aa 602-620) and a cytoplasmic tail (aa. 621-738 of isoform Long) that plays an essential role in PECAM-1-mediated cellular signaling. PECAM-1 is essential for leukocyte transendothelial migration (TEM) under most inflammatory conditions. It is expressed on platelets and leukocytes and is primarily concentrated at the borders between endothelial cells. Six isoforms of PECAM-1 have been described that are produced by alternative splicing. Isoform Long predominates in all tissues examined, whereas Delta 12 isoform is detected only in trachea and isoform Delta14-15 are only detected in lung tissue. Isoform Delta14 is detected in all tissues examined with the strongest expression in heart. Isoform Delta15 is expressed in brain, testis, ovary, cell surface of platelets, human umbilical vein endothelial cells (HUVECs), Jurkat T-cell leukemia, human erythroleukemia and U-937 histiocytic lymphoma cell lines. Tyrosine 690 residue plays a critical role in TEM and is required for efficient trafficking of PECAM-1 to and from the lateral border recycling compartment (LBRC) and is also required for the LBRC membrane to be targeted around migrating leukocytes. PECAM-1 also prevents phagocyte ingestion of closely apposed viable cells by transmitting ′detachment′ signals, and changes function on apoptosis, promoting tethering of dying cells to phagocytes.

Specificity

Clone MBC 78.1 detects CD31/PECAM-1 in human cells. It targets an epitope within the IgG homology domain 5.

Immunogen

PECAM-1 purified from human platelets.

Application

Anti-CD31 (PECAM-1), clone MBC 78.1, Cat. No. MABF2032, is a mouse monoclonal antibody that detects CD31/PECAM-1 and has been tested for use in Flow Cytometry, Immunocytochemistry, and Immunoprecipitation.
Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected CD31 (PECAM-1) in THP-1 cells.

Immunoprecipitation Analysis: A representative lot detected CD31 (PECAM-1) in Immunoprecipitation applications (DeLisser, H.M., et. al. (1994). J Cell Biol. 124(1-2):195-203).

Flow Cytometry Analysis: A representative lot detected CD31 (PECAM-1) in Flow Cytometry applications (DeLisser, H.M., et. al. (1994). J Cell Biol. 124(1-2):195-203).

Flow Cytometry Analysis: 1 µg from a representative lot detected CD31 (PECAM-1) in one millioin THP-1 cells.
Research Category
Inflammation & Immunology

Quality

Evaluated by Immunocytochemistry in Jurkat cells.

Immunocytochemistry Analysis: A 1:500 dilution of this antibody detected CD31 (PECAM-1) in Jurkat cells.

Target description

82.54 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service