Skip to Content
MilliporeSigma

MABF121

Anti-SV40 Large T Antigen Antibody, clone PAb416

clone PAb416, from mouse

Synonym(s):

Large T antigen, LT, LT-AG

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
PAb416, monoclonal
Application:
ICC, WB
Citations:
20
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

PAb416, monoclonal

species reactivity

SV40-infected cells

technique(s)

immunocytochemistry: suitable, western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

General description

Both the Simian virus (SV40) large T and small T antigens are encoded by the early region of the SV40 genome. The large T antigen binds DNA, and complexes with a 53,000 dalton cellular protein, p53, which is required for initiation of viral DNA replication during lytic growth. In addition the large T antigen binds DNA polymerase and the transcription factor AP-2 and forms a specific complex with the P105 product of the retinoblastoma susceptibility gene.
~82 kDa observed

Immunogen

Epitope: Large T antigen of SV40
Purified large T-antigen of SV40

Application

Anti-SV40 Large T Antigen Antibody, clone PAb416 is an antibody against SV40 Large T Antigen for use in Western Blotting, ICC.
Immunocytochemistry Analysis: A representative lot from an independent laboratory detected SV40 Large T Antigen in ICC (Sabatier, J., et al. (2005). 58(4):429-431.; Del Valle, L., et al. (2004). J Virol. 78(7):3462-3469.).
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Biochem/physiol Actions

This antibody recognizes the large T antigen of SV40.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blot in Cos-1 cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected SV40 Large T Antigen in 10 µg of Cos-1 cell lysate.
Control
Cos-1 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library