Skip to Content
MilliporeSigma

MABE971

Anti-HNF-1-beta Antibody, clone 12A5.1

clone 12A5.1, from mouse

Synonym(s):

Hepatocyte nuclear factor 1-beta, HNF-1B, HNF-1-beta, Homeoprotein LFB3, Transcription factor 2, TCF-2, Variant hepatic nuclear factor 1, vHNF1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
12A5.1, monoclonal
Application:
IHC, WB
Citations:
1
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

12A5.1, monoclonal

species reactivity

rat, human

technique(s)

immunohistochemistry: suitable, western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HNF1B(6928)

General description

Hepatocyte nuclear factor 1-beta (UniProt P35680; also known as HNF-1-beta, HNF-1B, Homeoprotein LFB3, TCF-2, Transcription factor 2, Variant hepatic nuclear factor 1, vHNF1) is encoded by the HNF1B (also known as TCF2, VHNF1) gene (Gene ID 6928) in human. HNF-1-beta is involved in transcriptional and functional regulation of the liver and biliary system, as well as of the kidneys, urogenital tract and pancreatic β-cells. HNF-1-beta deficiency due to HNF1B mutations is a well known cause of maturity-onset diabetes of the young 5 (MODY5), also referred to as RCAD (renal cysts and diabetes) due to the frequent co-occurrence of renal cysts and/or other inborn urogenital abnormalities. HNF-1-beta is expressed in the gall bladder, liver and intrahepatic bile ducts of mouse embryos and in liver of adult mice. HNF-1-beta is shown to co-regulate morphogenesis of the biliary system and HNF1B knockout mice suffer from severe neonatal cholestasis and jaundice due to the abnormally developed gall bladder and dysfunctional intrahepatic bile ducts.
~61 kDa observed

Immunogen

Epitope: Internal (N-terminal half)
GST-tagged recombinant protein corresponding to the Internal (N-terminal half) of human HNF-1-beta.

Application

Anti-HNF-1-beta Antibody, clone 12A5.1 is an antibody against HNF-1-beta for use in Western Blotting and Immunohistochemistry.
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected HNF1B in human kidney and rat liver tissue.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors

Biochem/physiol Actions

Expected to react with isoform 1 (A) and isoform 3 (C). Reactivity toward isoform 2 (B) and isoform 4 has not been determined.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in rat brain tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected HNF1B in 10 µg of rat brain tissue lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library