Skip to Content
MilliporeSigma
All Photos(2)

Documents

MABE912

Sigma-Aldrich

Anti-Ikaros Antibody, clone 4E9

clone 4E9, from mouse

Synonym(s):

DNA-binding protein Ikaros, Ikaros family zinc finger protein 1, Lymphoid transcription factor LyF-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

4E9, monoclonal

species reactivity

human, mouse

technique(s)

ChIP: suitable (ChIP-seq)
flow cytometry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... IKZF1(10320)
mouse ... Ikzf1(22778)

General description

Ikaros, also known as IKZF1, is a transcription factor that is important for the regulation of lymphopoiesis and also the development of the immune system. It has also been linked to adult and fetal erythroid differentiation in humans. Ikaros has been shown to induce local chromatin accessibility during CD4+ T cell differentiation and therefore, activating Th2 expression.

Immunogen

Recombinant protein corresponding to Human Ikaros.

Application

Flow Cytometry Analsysis: 1 µg of a representative lot of this antibody detected Ikaros in Mouse thymocytes

Chromatin Immunoprecipitation (ChIP): A representative lot of this antibody as part of a blend Ikaros clones E9 and Ik14 (catalogue number MABE912 and MABE913 respectively) was demonstrated to perform in ChIP. See Zhang et al. (2012).

ChIP-seq Analsysis: A representative lot of this antibody as part of a blend Ikaros clones E9 and Ik14 (catalogue number MABE912 and MABE913 respectively) was demonstrated to perform in ChIP-seq. See Zhang et al. (2012).
Research Category
Epigenetics & Nuclear Function

Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Chromatin Biology
This Anti-Ikaros Antibody, clone 4E9 is validated for use in Western Blotting and Flow Cytometry and Chromatin Immunoprecipitation (ChIP) and ChIP-seq for the detection of Ikaros.

Quality

Evaluated by Western Blotting in a Jurkat cell lysate.

Western Blotting Analysis: A 1 µg/mL dilution of this antibody detected Ikaros in a Jurkat cell lysate.

Target description

~60 kDa observed. Uncharacterized bands may be observed in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jingjie Chang et al.
Frontiers in immunology, 14, 1239779-1239779 (2023-09-04)
AIOLOS, encoded by IKZF3, is a member of the IKZF family of proteins that plays an important role in regulating late B-cell differentiation. Human individuals heterozygous for the AIOLOS p.N160S variant displayed impaired humoral immune responses as well as impaired

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service