Skip to Content
MilliporeSigma

MABE78

Anti-MiTF Antibody, clone 3D1.2

clone 3D1.2, from mouse

Synonym(s):

Microphthalmia-associated transcription factor, Class E basic helix-loop-helix protein 32, bHLHe32

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
3D1.2, monoclonal
Application:
IHC, WB
Citations:
4
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3D1.2, monoclonal

species reactivity

human, mouse

technique(s)

immunohistochemistry: suitable, western blot: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MITF(4286)

General description

Microphthalmia-associated transcription factor (MiTF) is a basic helix loop helix leucine zipper (b HLH ZIP) transcription factor implicated in pigmentation, and development of mast cells and bone. Mutations in MiTF cause auditory pigmentary syndromes in humans, such as Waardenburg syndrome type II and type IIa, and Tietz syndrome. There are two known isoforms of MiTF, differing by 66 amino acids at the N-terminus. Shorter forms are expressed in melanocytes and run as two bands at 52 kDa and 56 kDa, whereas the longer Mi form runs as a cluster of bands at 60-70 kDa in osteoclasts and in B16 melonoma cells (but not other melanoma cell lines), as well as mast cells and heart. MiTF plays a critical role in the differentiation of various cell types as neural crest-derived melanocytes, mast cells, osteoclasts and optic cup-derived retinal pigment epithelium.
~52 kDa observed. Multiple isoforms between 46-56 kDa are known to exist.

Immunogen

GST-tagged recombinant protein corresponding to human MiTF.

Application

Anti-MiTF Antibody, clone 3D1.2 is an antibody against MiTF for use in Western Blotting & IHC.
Immunohistochemistry Analysis: A 1:100 dilution of this antibody detected MiTF in human melanoma tissue.
Research Category
Signaling
Research Sub Category
Transcription Factors

Physical form

Format: Purified
Purified mouse monoclonal IgMκ in buffer containing 0.01M PBS, 0.1% sodium azide, pH 7.1.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse brain tissue lysate
Evaluated by Western Blot in mouse brain tissue lysate.

Western Blot Analysis: A 1:500 dilution of this antibody detected MiTF in 10 µg of mouse brain tissue lysate.

Other Notes

Replaces: MAB3747

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Shiyao Pei et al.
Experimental dermatology, 27(10), 1120-1125 (2018-07-26)
The long noncoding RNA H19 was reported to associate with melanogenesis. However, it remains unknown whether H19 expression will be changed by UVB irradiation and whether H19 will regulate melanocytes melanogenesis by paracrine effects. Here, we analysed the expression changes
Shiyao Pei et al.
The Journal of investigative dermatology, 140(1), 152-163 (2019-07-06)
The long noncoding RNA UCA1 was first discovered in bladder cancer and is known to regulate the proliferation and migration of melanoma. However, its role in melanogenesis is unclear. In this study, we aimed to explore the role and mechanism
Aishwarya Balasankar et al.
Frontiers in medicine, 9, 821361-821361 (2022-11-01)
Dental pulp stem cells (DPSCs) are an easily accessible, heterogenous source of mesenchymal stem cells (MSCs) that are derived from the neural crest. Evidence suggests that they have neurotrophic qualities in their undifferentiated state and can also be differentiated into



Global Trade Item Number

SKUGTIN
MABE7804053252698842