Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABE362

Sigma-Aldrich

Anti-EZH2 Antibody, clone AE25

clone AE25, from mouse

Synonym(s):

Enhancer of zeste homolog 2, Lysine N-methyltransferase 6, enhancer of zeste 2, histone-lysine N-methyltransferase EZH2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

AE25, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... EZH2(2146)

General description

EZH2 (enhancer of zeste homologue 2), also known as Enx1, is a member of the polycomb group of proteins and is involved in cell cycle regulation. EZH2 represses transcription via trimethylation of histone H3 on Lys27 (H3K27) as indicated by the fact that RNAi-mediated knockdown of EZH2 resulted in a loss of H3K27 trimethylation. EZH2 is ubiquitously expressed during early embryo genesis, and becomes restricted to the central and peripheral nervous systems and sites of fetal hematopoiesis during later development. In the adult, EZH2 is restricted to the spleen, testis, and placenta. EZH2 and BMI-1 genes are coexpressed in Reed-Sternberg cells of Hodgkin’s disease. Coexpression of BMI-1 and EZH2 is also associated with cycling cells and degree of malignancy in B-cell non-Hogkin’s lymphoma. EZH2 is involved in the progression of prostate cancer, and has been identified as a marker of aggressive breast cancer. Two transcript variants encoding distinct isoforms have been identified for this gene.

Immunogen

MBP-tagged recombinant protein corresponding to human EZH2.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology
This Anti-EZH2 Antibody, clone AE25 is validated for use in Western Blotting, IHC for the detection of EZH2.
Western Blot Analysis: A 1:500 dilution from a representative lot detected EZH2 in Jurkat and HeLa cell lysates.

Immunohistochemistry Analysis: A representative lot detected EZH2 in breast carcinoma tissue (Pietersen et al., 2008)

Quality

Evaluated by Western Blot in T47D cell lysate.

Western Blot Analysis: A 1:1000 dilution of this antibody detected EZH2 in 10 µg of T47D cell lysate.

Target description

~80 kDa observed. Uniprot describes multiple isoforms ranging from 80-86 kDa

Linkage

Replaces: 04-1047

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
T47D cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoning Wu et al.
The Prostate, 79(10), 1079-1089 (2019-05-20)
Enhancer of zeste 2 (EZH2) promotes prostate cancer progression. We hypothesized that increased EZH2 expression is associated with postradiotherapy metastatic disease recurrence, and may promote radioresistance. EZH2 expression was investigated using immunohistochemistry in diagnostic prostate biopsies of 113 prostate cancer
William A Scott et al.
PLoS genetics, 17(11), e1009909-e1009909 (2021-11-16)
The ATRX ATP-dependent chromatin remodelling/helicase protein associates with the DAXX histone chaperone to deposit histone H3.3 over repetitive DNA regions. Because ATRX-protein interactions impart functions, such as histone deposition, we used proximity-dependent biotinylation (BioID) to identify proximal associations for ATRX.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service