Skip to Content
MilliporeSigma

MABE251

Anti-5-hydroxymethylcytosine (5hmC) Antibody, clone HMC 31

clone HMC 31, from mouse

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
HMC 31, monoclonal
Technique(s):
ELISA: suitable, dot blot: suitable, methylated DNA immunoprecipitation (MeDIP): suitable
Application:
DB, ELISA, methylated DNA immunoprecipitation (MeDIP)
Citations:
3
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

HMC 31, monoclonal

species reactivity (predicted by homology)

all

technique(s)

ELISA: suitable, dot blot: suitable, methylated DNA immunoprecipitation (MeDIP): suitable

isotype

IgG1κ

shipped in

wet ice

target post-translational modification

unmodified

General description

5-Hydroxymethylcytosine is a DNA pyrimidine nitrogen base formed from the enzymatic conversion of 5-methylcytosine into 5-hydroxymethylcytosine by the TET family of iron-dependent oxygenases. Data suggests that every mammilian cell contains 5-hydroxymethylcytosine, but the levels vary depending on the cell type; data also suggests that levels of 5-hydroxymethylcytosine increases with age. The highest levels are found in neuronal cells of the central nervous system and certain mammalian tissues such as mouse Purkinje and granule neurons. Although the exact function has not been fully elucidated, studies suggest that 5-hydroxymethlcytosine may regulate gene expression or initiate DNA demethylation.
This antibody detects 5-hydroxymethylcytosine.

Immunogen

BSA-conjugated 5-hydroxymethylcytosine.
Epitope: 5-hydroxymethylcytosine

Application

Detect 5-hydroxymethylcytosine (5hmC) with Anti-5-hydroxymethylcytosine (5hmC) Antibody, clone HMC 31 (Mouse Monoclonal Antibody), that has been shown to work in DB, MeDIP, ELISA.
ELISA Analysis: A representative lot detected 5-hydroxymethylcytosine in ELISA.

Methylated DNA Immunoprecipitation (MeDIP) Analysis: A representative lot immunoprecipitated 5-hydroxymethylcytosine in ChIP.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Biochem/physiol Actions

This antibody recognizes the 5-hydroxymethylcytosine of 5-Hydroxymethylcytosine DNA.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Unmodified and modified cytosine DNA.
Evaluated by Dot Blot in unmodified and modified cytosine DNA.

Dot Blot Analysis: 0.1 µg/mL of this antibody detected 5-hydroxymethylcytosine in 10 ng and 50 ng of unmodified and modified cytosine DNA.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library