Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABE1328

Sigma-Aldrich

Anti-INK4a (p16) Antibody, clone 13H4.1

clone 13H4.1, from mouse

Synonym(s):

Cyclin-dependent kinase inhibitor 2A, CDK4I, Multiple tumor suppressor 1, MTS-1, p16-INK4, p16-INK4a, p16INK4A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

13H4.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... CDKN2A(1029)

General description

Cyclin-dependent kinase inhibitor 2A (UniProt P42771; also known as CDK4I, Cyclin-dependent kinase 4 inhibitor A, Multiple tumor suppressor 1, MTS-1, p16-INK4, p16-INK4a, p16INK4A) is encoded by the CDKN2A (also known as CDKN2, MTS1) gene (Gene ID 1029) in human. The INK and CIP/KIP families of cyclin dependent kinase inhibitors (CDKis) protect cells from oncogenic signals-initiated cellular transformation. INK4a and INK4b (encoded by the CDKN2A and CDKN2B gene, respectively) constitute the INK4 subfamily of CDKis. INK4a and INK4b are induced by oncogenic Ras and are involved in cell cycle G1 arrest and cellular senescence. Inactivation of CDKN2A and CDKN2B are found in a wide variety of human malignancies and murine tumor models.

Specificity

Clone 13H4.1 epitope lies within a region present in p16INK4a and the spliced isoforms 2 and 5 (UniProt P42771-1, P42771-2, and P42771-4 respectively), but absent from the spliced isoform 3 (p12; UniProt P42771-3), p14ARF/p19ARF or smARF (UniProt Q8N726-1 and Q8N726-2).

Immunogen

GST-tagged recombinant human p16INK4a internal fragment.

Application

Anti-INK4a (p16), clone 13H4.1 Antibody, Cat. No. MABE1451, is a highly specific mouse monoclonal antibody that targets INK4a (p16) and has been tested in Immunocytochemistry and Western Blotting.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected p16INK4a in human kidney and small intestine tissue sections.
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected p16INK4a in 10 µg of HEK293 cell lysate.

Target description

~16 kDa observed. 16.53 kDa (isoform 1; p16INK4a), 11.22 kDa (isoform 2), and 17.88 (isoform 5; p16gamma) calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified.
Purified mouse monoclonal IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4) 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Jin Sun Hwang et al.
International journal of molecular sciences, 21(12) (2020-06-25)
Human corneal endothelial cells (hCECs) pump out water from the stroma and maintain the clarity of the cornea. The sex-determining region Y-box 2 (SOX2) participates in differentiation during the development of the anterior segment of the eye and is found
null
Jin Sun Hwang et al.
Investigative ophthalmology & visual science, 61(4), 21-21 (2020-04-18)
Human corneal endothelial cells (hCECs) have limited regenerative capacity in vivo. Reduced hCEC density results in bullous keratopathy requiring corneal transplantation. This study reveals the role of transcription factor 4 (TCF4) in hCEC diseases and suggests that TCF4 may be
Zilong Yan et al.
Journal of experimental & clinical cancer research : CR, 38(1), 221-221 (2019-05-28)
Extracellular signal-regulated kinases (ERKs) have been related to multiple cancers, including breast cancer, hepatocellular cancer, lung cancer and colorectal cancer. ERK1/2 inhibitor can suppress growth of KRAS-mutant pancreatic tumors by targeting cancer cell. However, no studies have shown the expression

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service