Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
2H10, monoclonal
Application:
ICC, WB
Citations:
-
biological source
rat
Quality Segment
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
2H10, monoclonal
species reactivity
human, mouse, rat
technique(s)
immunocytochemistry: suitable, western blot: suitable
isotype
IgG2cκ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... NUP98(4928)
General description
The human gene NUP98 (Gene ID 4928; Also known as ADAR2, ADIR2, NUP96, NUP196) encodes the nuclear pore complex protein Nup98-Nup96 (UniProt P52948; also known as GLFG-repeat containing nucleoporin, Nup98-Nup96), which is posttranslationally autocleaved to form Nup98 and Nup96. Cleavage of the Nup98-96 linkage enhances the interaction between Nup98 and Nup96 and promotes localization of the two proteins to the nuclear pore complex (NPC). An alternative splice event generates a mRNA variant (isoform 3; also known as Nup98-specific 1), which expresses only the Nup98 portion of the Nup98-96 mRNA and allows the independent production of Nup98. Unlike Nup96, Nup98 is known to exist in both NPC-bound and non-NPC-associated pool, where it may participate in gene transcription regulations. Nup98 is reported to interact with histone-modifying enzymes CBP/p300 and histone deacetylases (HDACs) through its glycine-leucine-phenylalanine-glycine (GLFG) repeats. NUP98 gene is also involved in chromosomal translocations, resulting in Nup98 translocation fusions identified as potent transcription factors that induce acute myeloid leukemia (AML) with a particularly poor patient prognosis.
~100 kDa observed
Immunogen
Epitope: Near N-terminus
GST-tagged recombinant protein corresponding to human Nup98 near the N-terminus.
Application
Anti-Nup98 Antibody, clone 2H10 is an antibody against Nup98 for use in Western Blotting and Immunocytochemistry.
Immunocytochemistry Analysis: A representative lot detected Nup98 in Rat neurons.
Immunocytochemistry Analysis: A representative lot detected Nup98 nuclear localization in HeLa cells (Fukuhara, T., et al. (2005) Hybridoma (Larchmt). 24(5):244-247).
Immunocytochemistry Analysis: A representative lot detected nuclear localization Nup98 in NIH/3T3 cells (Oka, M., et al. (2010) Mol Biol Cell. 21(11):1885-1896).
Immunocytochemistry Analysis: A representative lot detected Nup98 nuclear localization in HeLa cells (Fukuhara, T., et al. (2005) Hybridoma (Larchmt). 24(5):244-247).
Immunocytochemistry Analysis: A representative lot detected nuclear localization Nup98 in NIH/3T3 cells (Oka, M., et al. (2010) Mol Biol Cell. 21(11):1885-1896).
Research Category
Epigenetics & Nuclear Function
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors
Nuclear Receptors
Biochem/physiol Actions
Reacts with Nup98-Nup96 precursor, nucleoprotein Nup98, but not nucleoprotein Nup96.
Physical form
Format: Purified
Protein G Purified
Purified rat monoclonal IgG2cκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Preparation Note
Stable for 1 year at 2-8°C from date of receipt.
Analysis Note
Evaluated by Western Blotting in HeLa nuclear extract.
Western Blotting Analysis: 1.0 µg/mL of this antibody detected Nup98 in 10 µg of HeLa nuclear extract.
Western Blotting Analysis: 1.0 µg/mL of this antibody detected Nup98 in 10 µg of HeLa nuclear extract.
Other Notes
Concentration: Please refer to lot specific datasheet.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.