Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

MABD42

Sigma-Aldrich

Anti-Cyp7a1 Antibody, clone 15B9.1

clone 15B9.1, from mouse

Synonym(s):

Cholesterol 7-alpha-monooxygenase, CYPVII, Cholesterol 7-alpha-hydroxylase, Cytochrome P450 7A1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

15B9.1, monoclonal

species reactivity

rat, mouse, human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CYP7A1(1581)

General description

Cytochrome P450 7A1 (CYP7A1, CYPVII), also known as cholesterol 7-alpha-monooxygenase or cholesterol 7-alpha-hydroxylase, belongs to the cytochrome P450 family and is important for cholesterol homeostasis. CYP7A1 catalyzes a rate-limiting step in cholesterol catabolism and bile acid biosynthesis by introducing a hydrophilic moiety at position 7 of cholesterol. Detected primarily in the liver, CYP7A1 catalyzes the following reaction: Cholesterol + NADPH + O2 = 7-alpha-hydroxycholesterol + NADP+ + H2O. CYP7A1 is up-regulated by glucose and by cholestyramine, and is down-regulated by chenodeoxycholic acid.

Immunogen

GST-tagged recombinant protein corresponding to human Cyp7a1.

Application

Imunnohistochemistry Analysis: A 1:50-1,000 dilution from a representative lot detected Cyp7a1 in rat and human liver tissue.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Cyp7a1 antibody is validated for use in WB & IHC for the detection of the Cyp7a1 protein.

Quality

Evaluated by Western Blotting in human liver tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Cyp7a1 in 10 µg of human liver tissue lysate.

Target description

~52 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human liver tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cen Xie et al.
Biochimica et biophysica acta. Molecular and cell biology of lipids, 1864(10), 1396-1411 (2019-06-14)
Peroxisome proliferator-activated receptor alpha (PPARα) controls lipid homeostasis through regulation of lipid transport and catabolism. PPARα activators are clinically used for hyperlipidemia treatment. The role of PPARα in bile acid (BA) homeostasis is beginning to emerge. Herein, Ppara-null and hepatocyte-specific
Hana Lastuvkova et al.
International journal of molecular sciences, 22(12) (2021-07-03)
Bile acids (BA) play a significant role in the pathophysiology of nonalcoholic steatohepatitis (NASH). The present study evaluates the modulation of bile acid metabolomics by atorvastatin, a cholesterol-lowering agent commonly used to treat cardiovascular complications accompanying NASH. NASH was induced
Sora Enya et al.
Disease models & mechanisms, 11(8) (2018-03-30)
The nature of host organs and genes that underlie tumor-induced physiological disruption on the host remains ill-defined. Here, we establish a novel zebrafish intestinal tumor model that is suitable for addressing this issue, and find that hepatic cyp7a1, the rate-limiting
Yongtao Xiao et al.
Cell death & disease, 8(10), e3110-e3110 (2017-10-13)
The p38α mitogen-activated protein kinase (MAPK) has been related to gluconeogenesis and lipid metabolism. However, the roles and related mechanisms of p38α MAPK in intestinal failure (IF)-associated liver steatosis remained poor understood. Here, our experimental evidence suggested that p38α MAPK
Michelle M Chen et al.
The Journal of endocrinology, 237(2), 139-152 (2018-04-05)
Fibroblast growth factor 21 (FGF21) is a potent regulator of glucose and lipid homeostasis in vivo; its most closely related subfamily member, FGF19, is known to be a critical negative regulator of bile acid synthesis. To delineate whether FGF21 also

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service