Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABC965

Sigma-Aldrich

Anti-Prohibitin-1 (PHB1), clone 1G5C1 Antibody

clone 1G5C1, from rat

Synonym(s):

Prohibitin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

1G5C1, monoclonal

species reactivity

human, mouse, rat

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PHB(5245)

General description

Prohibitin is an inhibitor of DNA synthesis that is widely expressed across various tissues. Prohibitin influences proliferation, and may also have a role in regulating mitochondrial respiration activity and in aging. Expression levels of prohibitin in fibroblasts have been found to decrease heterogeneously as cellular aging occurs.

Immunogen

KLH-conjugated linear peptide corresponding to human Prohibitin-1 (PHB1).

Application

Anti-Prohibitin-1 (PHB1), clone 1G5C1 | MABC965 is an antibody against Prohibitin-1 for use in Western Blotting, Immunohistochemistry.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected Prohibitin-1 (PHB1) in 10 µg of NIH/3T3 and HeLa cell lysate.
Immunohistochemistry Analysis: A representative lot detected Prohibitin-1 (PHB1) in rat liver tissue (Kakehashi, A., et al. (2011). Toxicological Sciences. 119(1):61-72).

Quality

Evaluated by Western Blotting in rat liver tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Prohibitin-1 (PHB1) in 10 µg of rat liver tissue lysate.

Target description

~30 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service