Skip to Content
MilliporeSigma

Skip To

MABC953

Anti-Prohibitin-2/PHB2 Antibody, clone 7F8E3

clone 7F8E3, from rat

Synonym(s):

Prohibitin-2, B-cell receptor-associated protein BAP37, D-prohibitin, Repressor of estrogen receptor activity, Prohibitin-2/PHB2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
7F8E3, monoclonal
Application:
ICC, IHC, WB
Citations:
1
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rat

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7F8E3, monoclonal

species reactivity

rat, monkey, human, mouse

technique(s)

immunocytochemistry: suitable, immunohistochemistry: suitable (paraffin), western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PHB2(11331)

General description

Prohibitin-2 (UniProt Q99623; also known as B-cell associated protein, B-cell receptor-associated protein BAP37, BCAP37, D-prohibitin, PHB2, Repressor of estrogen receptor activity) is encoded by the PHB2 (also known as BAP, REA) gene (Gene ID 11331) in human. Prohibitins are assembled into a ring-like PHB complex with 16-20 alternating Phb1 and Phb2 subunits in the inner mitochondrial membrane, where they function as chaperone for respiration chain proteins or as important structural scaffold for optimal mitochondrial morphology and function. Prohibitins are also reported to modulate transcriptions by interacting with various transcription factors, including nuclear receptors. Upregulated expression of prohibitins, septin 9, neurabin 1, and other cytoskeletal and functional proteins is found at the glutathione S-transferase placental-form-positive (GST-P+) hepatocellular carcinoma (HCC) foci during initiation and/or promotion stages of rat hepatocarcinogenesis, implicating an involvement of prohibitins in the neoplastic transformation of rat liver preneoplastic lesions.
~34 kDa observed

Immunogen

Epitope: Near C-terminus
KLH-conjugated linear peptide corresponding to human Prohibitin-2/PHB2.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-Prohibitin-2/PHB2 Antibody, clone 7F8E3 is validated for use in Western Blotting, Immunocytochemistry, Immunohistochemistry (Paraffin) for the detection of Prohibitin-2/PHB2.
Western Blotting Analysis: 0.5 µg/mL from a representative lot detected Prohibitin-2/PHB2 in 10 µg of COS-7 cell lysate.
Immunocytochemistry Analysis: 4 µg/mL from a representative lot detected Prohibitin-2/PHB2 in A431 and HUVEC cells.
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected Prohibitin-2/PHB2 in human breast and mouse liver tissue.
Immunohistochemistry Analysis: A representative lot detected upregulated Prohibitin-2 immunoreactivity at the glutathione S-transferase placental-form–positive (GST-P+) hepatocellular carcinoma (HCC) foci in frozen liver sections from rats subjected to hepatocarcinogenesis induction by N-nitrosodiethylamine (DEN) and Phenobarbital (PB) treatment (Kakehashi, A., et al. (2011). Toxicol Sci. 119(1):61-72).

Biochem/physiol Actions

Expected to react with both spliced isoforms.

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2aκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in rat C6 glioma cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Prohibitin-2/PHB2 in 10 µg of rat C6 glioma cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB5701681HPA039874ABN293
antibody form

purified immunoglobulin

antibody form

-

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

species reactivity

rat, monkey, human, mouse

species reactivity

human, rat, mouse

species reactivity

human, rat, mouse

species reactivity

human

Gene Information

human ... PHB2(11331)

Gene Information

human ... PHB2(11331)

Gene Information

human ... PHB2(11331)

Gene Information

human ... PHB(5245)

clone

7F8E3, monoclonal

clone

monoclonal

clone

polyclonal

clone

polyclonal

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

biological source

rat

biological source

rabbit

biological source

rabbit

biological source

rabbit


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters