Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MABC1766

Sigma-Aldrich

Anti-LAMP2/CD107b Antibody, clone H4B4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

H4B4, monoclonal

mol wt

calculated mol wt 44.96 kDa
observed mol wt ~120 kDa

purified by

using protein G

species reactivity

mouse, rat, human, monkey

packaging

antibody small pack of 100 μL

technique(s)

ELISA: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... LAMP2(3920)

General description

Lysosome-associated membrane glycoprotein 2 (UniProt: P13473; also known as LAMP-2, Lysosome-associated membrane protein 2, CD107 antigen-like family member B, LGP-96, CD107b) is encoded by the LAMP2 gene (Gene ID: 3920) in human. LAMP-2 is a single-pass type I, heavily glycosylated, membrane protein found in plasma membrane, endosomes, and lysosomes. It is synthesized with a signal peptide (aa 1-28), which is subsequently cleaved off to generate the mature form that contains a lumenal domain (aa 29-375), a transmembrane domain (aa 376-399), and a short cytoplasmic tail (aa 400-410). It is an important component of lysosomal membranes and is involved in both autophagy and lysosomal membrane permeabilization. LAMP-2 plays an important role in chaperone-mediated autophagy. It is required for the fusion of autophagosomes with lysosomes during autophagy. It also plays a role in lysosomal protein degradation in response to starvation. It functions by binding target proteins, such as GAPDH and MLLT11, and targets them for lysosomal degradation. In cancer cells, chronic acidosis-induced upregulation of LAMP-2 has been reported that protects these cells from acid-induced hydrolysis and promotes their survival via chaperone-mediated autophagy. Loss of LAMP-2 is reported to aggravate oxidative stress through the obstruction of reactive oxygen species clearance. Three different isoforms of LAMP-2 have been described that are produced by alternative splicing. Mutations in LAMP2 gene have been linked to Danon disease that is characterized by cardiomyopathy, vascular myopathy, and mental retardation. It is often associated with accumulation of glycogen in muscle and lysosomes.

Specificity

Clone H4B4 is a mouse monoclonal antibody that detects LAMP-2 (CD107b).

Immunogen

Human adherent peripheral blood cells

Application

Quality Control Testing

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected LAMP2/CD107b in Jurkat cell lysate.

Tested Applications

Western Blotting Analysis: A 1:500 dilution from a representative lot detected LAMP2/CD107b in HeLa, HepG2, and U937 cell lysates.

Western Blotting Analysis: A representative lot detected LAMP2/CD107b in Western Blotting applications (Krag, T.O., et al. (2021). Acta Neuropathol Commun. 9(1):109; Bozic, M., et al. (2020). EMBO Rep. 21(3):e48412; Shakya, S., et al. (2018). EMBO Rep. 19(12):e45918).

Immunohistochemistry Analysis: A representative lot detected LAMP2/CD107b in Immunohistochemistry applications (Krag, T.O., et al. (2021). Acta Neuropathol Commun. 9(1):109; Orenstein, S.J., et al. (2013). Nat Neurosci. 16(4):394-406).

Immunoprecipitation Analysis: A representative lot immunoprecipitated LAMP2/CD107b in Immunoprecipitation applications (Li, C., et al. (2017). EMBO J. 36(4):441-457).

Flow Cytometry Analysis: 1 μg from a representative lot detected LAMP2/CD107b in one million HEK293 cells.

ELISA Analysis: A representative lot detected LAMP2/CD107b in ELISA applications (Willy, J.A., et al. (2017). J Biol Chem. 292(34):14050-14065).

Immunofluorescence Analysis: A representative lot detected LAMP2/CD107b in Immunofluorescence applications (Gudi, R., et. al. (2021). Sci Rep. 11(1):12689; Krag, T.O., et al. (2021). Acta Neuropathol Commun. 9(1):109; Orenstein, S.J., et al. (2013). Nat Neurosci.;16(4):394-406; Tam, J., et al. (2014). PLoS One. 9(7):e101772).

Flow Cytometry Analysis: A representative lot detected LAMP2/CD107b in Flow Cytometry applications (Kornicka, K., et al. (2019). J Cell Mol Med. 23(1):237-259; Bhattacharya, S., et al. (2017). Invest Ophthalmol Vis Sci. 58(4):2366-2387).

Immunocytochemistry Analysis: A representative lot detected LAMP2/CD107b in Immunohistochemistry applications (Gudi, R., et. al. (2021). Sci Rep. 11(1):12689; Allen, E.A., et al. (2020). J Cell Biol. 219(11):e201909073; Tam, J., et al. (2014). PLoS One. 9(7):e101772).

Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution from a representative lot detected LAMP2/CD107b in human liver tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-LAMP2/CD107b, clone H4B4, Cat. No. MABC1766, is a mouse monoclonal antibody that detects LAMP2 and is tested in ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG1 in PBS without azide.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service