Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABC1637

Sigma-Aldrich

Anti-xCT Antibody, clone 3A12

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.74

biological source

rat

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

3A12, monoclonal

mol wt

calculated mol wt 55.42 kDa
observed mol wt ~57 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100 μg

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG2a

epitope sequence

N-terminal cytoplasmic domain

Protein ID accession no.

UniProt accession no.

shipped in

2-8°C

target post-translational modification

unmodified

Gene Information

General description

Cystine/glutamate transporter (UniProt: Q9UPY5; also known as Amino acid transport system xc-, Calcium channel blocker resistance protein CCBR1, Solute carrier family 7 member 11, xCT) is encoded by the SLC7A11 gene (Gene ID: 23657) in human. xCT is a multi-pass membrane protein with seven cytoplasmic domains, twelve transmembrane domains, and six extracellular domains. It serves as a sodium-independent, high-affinity exchange of anionic amino acids and has high specificity for anionic form of cystine and glutamate. Its expression is induced in response to reactive oxygen species (ROS)-inducing agents such as hydrogen peroxide, leading to enhanced glutathione (GSH) production. Higher expression of xCT is reported in astrocytes where it acts as an antiporter that couples the uptake of one molecule of cystine with the export of one molecule of glutamate. It is overexpressed in several cancers, including non-small cell lung cancer, breast cancer, and liver cancer and its elevated levels are associated with poor outcome. It has also been implicated in promoting tumorigenesis through its antioxidant function that supports cancer cell proliferation, matrix invasion, and tumor growth. Normally, KRAS is shown to protect fibroblasts against oxidative damage by stimulating xCT to enhance glutathione levels. However, in cancer cells xCT expression and activity can be induced by mutant KRAS, which can favor tumor growth. (Ref.: Lim, JKM., et al. (2019). Proc. Natl. Acad. Sci. USA. 116(19); 9433-9442).

Specificity

Clone 3A12 is a rat monoclonal antibody that detects human Cystine/glutamate transporter (SLC7A11). It targets an epitope within 21 amino acids from the N-terminal, first cytoplasmic domain.

Immunogen

Ovalbumin-conjugated linear peptide corresponding to 21 amino acids from the N-terminal, first cytoplasmic domain of human Cystine/glutamate transporter (SLC7A11).

Application

Quality Control Testing

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: A 1:125 dilution of this antibody detected Cystine/glutamate transporter (xCT) in HepG2 cell lysates.

Tested Applications

Western Blotting Analysis: A 1:125 dilution from a representative lot detected Cystine/glutamate transporter (xCT) in A549 cell lysate.

Immunocytochemistry Analysis: A 1:50 dilution from a representative lot detected V (xCT) in A549 cells.

Western Blotting Analysis: A representative lot detected Cystine/glutamate transporter (xCT) in Western Blotting applications (Doll, S., et al. (2019). Nature.;575(7784):693-698).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-xCT, clone 3A12, Cat. No. MABC1637, is a rat monoclonal antibody that detects Cystine/glutamate transporter (xCT) and is tested for use in Immunocytochemistry and Western Blotting.

Physical form

Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service