Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABC1635

Sigma-Aldrich

Anti-MLKL Antibody, clone 10C2

Synonym(s):

Mixed lineage kinase domain-like protein;hMLKL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rat

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

10C2, monoclonal

mol wt

calculated mol wt 54.78 kDa
observed mol wt ~50 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100 μL

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... MLKL(197259)

General description

Mixed lineage kinase domain-like protein (UniProt: Q8NB16; also known as hMLKL) is encoded by the MLKL gene (Gene ID: 197259) in human. MLKL is a pseudokinase that is a terminal protein in the pro-inflammatory necroptotic cell death program. It plays a critical role in TNF induced necroptosis. Although it contains a protein kinase domain (aa 194-469), it lacks several residues that are essential for protein kinase activity. It is activated following phosphorylation at Thr357/Ser358 by RIPK3, which leads to its homotrimerization and localization to the plasma membrane where it binds to highly phosphorylated inositol (InsP6) and cardiolipin and disrupts membrane integrity to cause necrotic cell death. Under basal conditions, MLKL is shown to reside in small puncta that are distributed evenly throughout the cytoplasm, but it is detected in plasma membrane shortly after its phosphorylation. It contains two coiled coil regions (aa 55-84 and 139-180). The second coiled coil region is responsible for its oligomerization. MLKL is also reported to be involved in endosomal trafficking, independently of induction of necroptosis, whereby it enhances degradation of receptors and ligands. Two isoforms of MLKL have been described that are produced by alternative splicing. (Ref.: Samson, AL., et al. (2020). Nat. Commun. 11(1); 3151; Yoon, S., et al. (2017). Immunity. 47(1); 51-65; Wang, H., et al (2014). Mol. Cell 54(1); 133-146).

Specificity

Clone 10C2 is a rat monoclonal antibody that detects human Mixed lineage kinase domain-like protein (MLKL). It targets an epitope within the C-terminal region.

Immunogen

GST-tagged recombinant fragment corresponding to 282 amino acids from human Mixed lineage kinase domain-like protein (MLKL) expressed in baculovirus expression system.

Application

Quality Control Testing

Evaluated by Western Blotting in wild-type HT-29 cell lysate.

Western Blotting Analysis (WB): A 1:500 dilution of this antibody detected MLKL in lysate from wild-type HT-29 cells, but not in MLKL knockout HT-29 cells.

Tested Applications

Immunocytochemistry Analysis: A representative lot detected MLKL in Immunocytochemistry applications (Samson, A.L., et al. (2020). Nat Commun. 11(1):3151).

Immunofluorescence Analysis: A representative lot detected MLKL in Immunofluorescence applications (Samson, A.L., et al. (2020). Nat Commun. 11(1):3151).

Western Blotting Analysis: A representative lot detected MLKL in Western Blotting applications (Samson, A.L., et al. (2020). Nat Commun. 11(1):3151).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-MLKL, clone 10C2, Cat. No. MABC1635, is a rat monoclonal antibody that detects MLKL and is tested for use in Immunocytochemistry, Immunofluorescence, and Western Blotting.

Physical form

Purified rat monoclonal antibodyIgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service